Skip to Content
Merck
All Photos(1)

Documents

O4755

Supelco

Oxantel pamoate

analytical standard

Synonym(s):

1-Methyl-2-(3-hydroxyphenylethenyl)-1,4,5,6-tetrahydropyrimidine compound with 4,4′-methylenebis(3-hydroxy-2-naphthoic acid), Oxantel embonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O · C23H16O6
CAS Number:
Molecular Weight:
604.65
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CN1CCCN=C1\C=C\c2cccc(O)c2.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(O)=O)c3O

InChI

1S/C23H16O6.C13H16N2O/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-15-9-3-8-14-13(15)7-6-11-4-2-5-12(16)10-11/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2,4-7,10,16H,3,8-9H2,1H3/b;7-6+

InChI key

CCOAINFUFGBHBA-UETGHTDLSA-N

General description

Oxantel pamoate is a drug used in the treatment of trichuriasis which is highly effective against the helminth of Trichuris sp.

Application

Oxantel pamoate may be used as a reference standard for examining the quality of pharmaceutical dosage forms containing oxantel pamoate (OPA) by reversed-phase high-performance liquid chromatography method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intestinal protozoan and helminth infections and control of soil-transmitted helminths in Malay school children.
B Sinniah
Public health, 98(3), 152-156 (1984-05-01)
Beatrice Barda et al.
The Lancet. Infectious diseases, 18(8), 864-873 (2018-06-03)
The recommended anthelmintics show low efficacy in a single-dose regimen against Trichuris trichiura. Moxidectin, a new treatment for river blindness, might complement the drug armamentarium for the treatment and control of soil-transmitted helminthiasis. However, its efficacy against T trichiura has
Development of oxantel tablets for pediatric clinical studies: a technical note.
Alles R, et al.
Journal of drug delivery science and technology, 23(6), 623-625 (2013)
K Peldán et al.
Scandinavian journal of infectious diseases, 14(4), 297-299 (1982-01-01)
166 patients with trichuriasis have been treated at the Department of Tropical Diseases, Aurora Hospital, Helsinki, from January 1975 to October 1979. Final results could be confirmed in 141 patients. Thiabendazole was found discouraging, because of frequent side effects and
Wendelin Moser et al.
The Lancet. Infectious diseases, 18(7), 729-737 (2018-04-21)
Albendazole and mebendazole are commonly used to control hookworm, but have shortcomings in their efficacy profiles. We assessed whether triple drug therapy (TDT) with albendazole, pyrantel pamoate, and oxantel pamoate was more effective than the co-administration of two drugs for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service