Skip to Content
Merck
All Photos(1)

Key Documents

NIST1543

GC/MS system performance

NIST® SRM® 1543

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 4 x 1 mL

manufacturer/tradename

NIST®

technique(s)

GC/MS: suitable

application(s)

general analytical

format

multi-component solution

General description

This Standard Reference Material (SRM) is intended primarily for use in evaluating the sensitivity of gas hromatography/mass spectrometry (GC/MS) instrumentation. A unit of SRM 1543 consists of four solutions: two concentrations of methyl stearate in hexane and two concentrations of benzophenone in hexane. For more information, please refer to the SDS and COA.

SRM 1543_cert

SRM 1543_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Methyl Stearate (Low) (C19H38O2), Methyl Stearate (High) (C19H38O2), Benzophenone (Low) (C13H10O), Benzophenone (High) (C13H10O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service