Skip to Content
Merck
All Photos(1)

Key Documents

G7391

Supelco

β-D-Glucan from barley

for use with Total Dietary Fiber Control Kit, TDF-C10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21

form

lyophilized powder

Quality Level

quality

for use with Total Dietary Fiber Control Kit, TDF-C10

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

storage temp.

2-8°C

InChI

1S/C18H32O16/c19-1-4-7(22)8(23)12(27)17(31-4)34-15-6(3-21)32-18(13(28)10(15)25)33-14-5(2-20)30-16(29)11(26)9(14)24/h4-29H,1-3H2/t4-,5-,6-,7-,8+,9-,10-,11-,12-,13-,14?,15?,16+,17+,18+/m1/s1

InChI key

FYGDTMLNYKFZSV-URKRLVJHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-D-Glucan is a β-D-glucose polysaccharide, naturally found in the cell wall of fungi, bacteria and plants.

Application

Internal control for use with the Total Dietary Fiber Assay Kit.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasma (1? 3)-beta-D-glucan measurement in diagnosis of invasive deep mycosis and fungal febrile episodes.
Obayashi T, et al.
Lancet, 345(8941), 17-20 (1995)
Overview of (1? 3)-beta-D-glucan immunobiology.
Williams LD
Mediators of Inflammation, 6(4), 247-250 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service