Skip to Content
Merck
All Photos(1)

Key Documents

900631

Sigma-Aldrich

Ethylene glycol

anhydrous, ZerO2®, 99.8%

Synonym(s):

Degassed and low oxygen ethylene glycol, Ethylene glycol ZerO2®, 1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
EC Number:
MDL number:
UNSPSC Code:
12352137
NACRES:
NA.21
grade:
ZerO2®
Assay:
99.8%
bp:
195-198 °C
vapor pressure:
0.08 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ZerO2®

Quality Level

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

99.8%

form

liquid

autoignition temp.

752 °F

quality

anhydrous

expl. lim.

15.3 %

impurities

≤0.003% water
water

refractive index

n20/D 1.431 (lit.)

bp

195-198 °C

mp

−13 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service