77396
Total Coliform ID Membrane
pink colored, sterile, diameter 70 mm, suitable for microbiology
Synonym(s):
Indicator membrane, ID Membranes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41171621
NACRES:
NA.85
Recommended Products
membrane size
70 mm
Quality Level
sterility
sterile
form
disc
shelf life
limited shelf life, expiry date on the label
storage condition
protect from light
technique(s)
microbe id | specific enzyme detection: suitable
application(s)
agriculture
environmental
food and beverages
microbiology
storage temp.
2-8°C
suitability
Escherichia coli
coliforms
General description
Total Coliform ID Membrane is a substrate-carrying membrane used for the rapid and qualitative detection of coliform bacteria in food and environmental samples. The discs are placed on the agar surface post-incubation and observed for characteristic color reactions. The membranes contain chromogenic substrates, which serve as the basis for microbe differentiation by color. The target organisms possess enzyme systems that metabolize the substrates in the membrane, leading to a change of color. The colors can be visually detected on the membrane. The principle is based on a specific substrate enzyme reaction in which coliform organisms will form blue, red, and pink-colored colonies.
Application
Total Coliform ID Membranes are substrate-carrying membranes used for the rapid and qualitative detection of coliform bacteria in food and environmental samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.
Analysis Note
Escherichia coli (25922) - Dark blueKlebsiella pneumoniae (13883) - Light pinkEnterobacter cloacae (23355) - Salmon to redCitrobacter freundii (8090) - Salmon to red
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service