Skip to Content
Merck
All Photos(1)

Documents

42157

Supelco

Procyanidin B2

analytical standard

Synonym(s):

(−)-Epicatechin (4β-8)-(−)-epicatechin, 4,8″-Bi-[(+)-epicatechin], cis,cis″-4,8″-Bi(3,3′,4′,5,7-pentahydroxyflavane), Proanthocyanidin B2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H26O12
CAS Number:
Molecular Weight:
578.52
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1Cc2c(O)cc(O)c([C@@H]3[C@@H](O)[C@H](Oc4cc(O)cc(O)c34)c5ccc(O)c(O)c5)c2O[C@@H]1c6ccc(O)c(O)c6

InChI

1S/C30H26O12/c31-13-7-20(37)24-23(8-13)41-29(12-2-4-16(33)19(36)6-12)27(40)26(24)25-21(38)10-17(34)14-9-22(39)28(42-30(14)25)11-1-3-15(32)18(35)5-11/h1-8,10,22,26-29,31-40H,9H2/t22-,26-,27-,28-,29-/m1/s1

InChI key

XFZJEEAOWLFHDH-NFJBMHMQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Procyanidin B2 is a B type of proanthocyanidin, that is present in plants, especially in grape seeds, apples and cacao seeds, which is known to possess anti-inflammatory properties and antioxidant properties.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier I Ottaviani et al.
The American journal of clinical nutrition, 95(4), 851-858 (2012-03-02)
Accumulating data show a causal role for flavanols in the mediation of cardiovascular benefits associated with the consumption of flavanol- and procyanidin-containing foods. Evidence for a direct causal role for procyanidins in this context is far less profound due to
Stavroula Stoupi et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(2), 287-291 (2009-11-17)
Procyanidins are important biologically active compounds, but the pathway and extent of absorption and metabolism are controversial. We conducted a mass balance study to evaluate the total radioactivity excreted in urine and feces after oral administration of [(14)C]procyanidin B2 to
Stavroula Stoupi et al.
Molecular nutrition & food research, 54(6), 747-759 (2009-11-28)
The catabolism by human faecal microbiota of (-)-epicatechin (1) (2, 3-cis stereochemistry) and its dimer pure procyanidin B2 (2), has been compared using a static in vitro culture model. The catabolites were characterised by LC-MS(n), UV absorption and relative retention
Nabeelah Bibi Sadeer et al.
Journal of pharmaceutical and biomedical analysis, 174, 19-33 (2019-06-04)
Africa is famous for its floral biodiversity, exploited by local people for therapeutic purposes. However, such plants need to be scrutinised scientifically for the presence of bioactive compounds and possible biological properties. This study attempts for the first time to
Absorption and urinary excretion of procyanidin B2 [epicatechin-(4?-8)-epicatechin] in rats
Baba S, et al.
Free Radical Biology & Medicine, 33, 142-148 (2012)

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service