Skip to Content
Merck
All Photos(1)

Key Documents

36707

Supelco

1,2-Dichlorobenzene

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein:
606078
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.04

grade

analytical standard

Quality Level

vapor density

5.1 (vs air)

vapor pressure

1.2 mmHg ( 20 °C)
1.6 mmHg ( 35 °C)

product line

PESTANAL®

autoignition temp.

1198 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

9.2 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.551 (lit.)

bp

178-180 °C (lit.)

mp

−18-−17 °C (lit.)

density

1.306 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccccc1Cl

InChI

1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

RFFLAFLAYFXFSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bimalendu Adhikari et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(41), 11488-11496 (2011-09-29)
The N-terminally pyrene-conjugated oligopeptide, Py-Phe-Phe-Ala-OMe, (Py=pyrene 1-butyryl acyl) forms transparent, stable, supramolecular fluorescent organogels in various organic solvents. One of these organogels was thoroughly studied using various techniques including transmission electron microscopy (TEM), field-emission scanning electron microscopy (FESEM), atomic force
Alexander S Filatov et al.
Inorganic chemistry, 49(4), 1626-1633 (2010-01-14)
The first example of a reversible [Cu(4)] <--> [Cu(6)] interconversion for polynuclear copper(I) complexes under controlled experimental settings is reported. It illustrates the key role of specific crystal growth conditions for accessing the target cluster nuclearity that consequently determines physical
Sungmin Chin et al.
Chemosphere, 75(9), 1206-1209 (2009-03-10)
This study examined the catalytic oxidation of 1,2-dichlorobenzene on V(2)O(5)/TiO(2) nanoparticles. The V(2)O(5)/TiO(2) nanoparticles were synthesized by the thermal decomposition of vanadium oxytripropoxide and titanium tetraisopropoxide. The effects of the synthesis conditions, such as the synthesis temperature and precursor heating
Eric Vejerano et al.
Environmental science & technology, 45(2), 589-594 (2010-12-09)
Previous studies have shown environmentally persistent free radicals (EPFRs) form when chlorine- and hydroxy-substituted benzenes chemisorb on Cu(II)O-containing surfaces under postcombustion conditions. This paper reports the formation of EPFRs on silica particles containing 5% Fe(III)(2)O(3). The EPFRs are formed by
Ivan Guryanov et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(46), 12837-12845 (2009-10-23)
The effect of microwave (MW) irradiation and ionic liquids (IL) on the cycloaddition of azomethine ylides to [60]fullerene has been investigated by screening the reaction protocol with regard to the IL medium composition, the applied MW power, and the simultaneous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service