Skip to Content
Merck
All Photos(1)

Key Documents

01-6140

Sigma-Aldrich

Antimony(III) chloride

JIS special grade, ≥98.0%

Synonym(s):

Antimony trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbCl3
CAS Number:
Molecular Weight:
228.12
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

vapor density

7.9 (vs air)

vapor pressure

1 mmHg ( 49 °C)

Assay

≥98.0%

form

crystalline powder

reaction suitability

reagent type: catalyst
core: antimony

availability

available only in Japan

mp

73.4 °C (lit.)

SMILES string

Cl[Sb](Cl)Cl

InChI

1S/3ClH.Sb/h3*1H;/q;;;+3/p-3

InChI key

FAPDDOBMIUGHIN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The response of different photometers to the color produced by vitamin A and carotene with antimony trichloride.
M J CALDWELL et al.
Transactions of the Kansas Academy of Science. Kansas Academy of Science, 49(2), 197-204 (1946-09-01)
S A Hussain et al.
Human & experimental toxicology, 17(3), 140-143 (1998-05-20)
1. The toxic gas hypothesis proposes exposure to stibine (antimony trihydride) generated from microbial contamination of cot mattress materials as a possible cause of unexplained death in infancy (SIDS) as a consequence of cholinesterase inhibition. We have measured the direct
N Schaumlöffel et al.
Mutagenesis, 13(3), 281-286 (1998-06-27)
Data on the mechanism of antimony genotoxicity is scarce. Arsenic and antimony are proposed to share some toxicological features. Thus comparative and combined experiments with As(III) and Sb(III) were performed to gain a deeper knowledge of the mechanism of antimony
Why not Nichols chemosurgery?
Gary A Dyer
Clinics in dermatology, 24(5), 458-460 (2006-09-13)
T Gebel
Mutation research, 412(3), 213-218 (1998-05-26)
Arsenic and antimony are two semimetals sharing some chemical as well as toxicological properties. Both elements are clastogenic but not point mutagenic in their trivalent state of valency. Environmental exposure to arsenic was proven to be associated with increased rates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service