Skip to Content
MilliporeSigma

Skip To

MX0214

Mannitol

Meets ACS Specifications GR ACS

Synonym(s):

D-Mannitol, D-Mannitol, Mannite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
3 kg
Check Cart for Availability
$354.00

About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
EC Index Number:
200-711-8
UNSPSC Code:
12352201
Beilstein/REAXYS Number:
1721898
MDL number:

$354.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

color

white

pH

5-7 (20 °C, 100 g/L in H2O)

bp

290-295 °C/4 hPa

mp

167-170 °C (lit.)

solubility

213 g/L

shipped in

ambient

storage temp.

room temp

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Application

Meets ACS Specifications GR ACS

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Analysis Note

ColorWhiteFormPowderHeavy metals (as Pb)5ppm maxInsoluble matter0.01% maxLoss on drying (105°C)0.05% maxReducing sugarsTo pass testResidue after ignition0.01% maxSpecific rotation [a]25/D+23.3-+24.3°Titrable acid0.0008meq/g max

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1007M0200000M8429
solubility

213 g/L

solubility

-

solubility

-

solubility

water: soluble 182 g/L at 20 °C (68 °F)

form

powder

form

-

form

-

form

powder

storage temp.

room temp

storage temp.

2-30°C

storage temp.

-

storage temp.

-

mp

167-170 °C (lit.)

mp

167-170 °C (lit.)

mp

167-170 °C (lit.)

mp

167-170 °C (lit.)

color

white

color

-

color

-

color

white

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-


Still not finding the right product?

Explore all of our products under Mannitol


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters