Skip to Content
Merck
All Photos(1)

Key Documents

MABN481

Sigma-Aldrich

Anti-Lipocalin-2 Antibody, clone PA348-26.3.5

clone PA348-26.3.5, from mouse

Synonym(s):

ngal, neutrophil lipocalin, siderocalin, Neutrophil gelatinase-associated lipocalin, NGAL, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, Lipocalin-2, Oncogene 24p3, Siderocalin LCN2, p25

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

PA348-26.3.5, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LCN2(3934)

General description

Lipocalin-2 (LCN2) (also known as Neurophil Gelatinase-Associated Lipocalin or NGAL) belongs to a superfamily of lipocalins, a collection of small, diversely functional, extracellular proteins. Lipocalin-2 is secreted from neutrophil granules in three different forms: a monomer of 25 kDa, a disulfide-linked homodimer at 46 kDa, and a disulfide-linked heterodimer with MMP-9 at 135-kDa. It is widely known to induce development of kidney epithelia, to bind and traffic iron, and to act as a molecular signal under inflammatory conditions, such as Crohn’s Disease. Multiple carcinoma studies have also shown elevated expression levels of Lipocalin-2 in several different human tumors affecting such organs as colon, lung, breast, ovaries, and pancreas. Additionally, there has been great interest in this protein as a possible marker for onset of Acute Kidney Injury following cardiac surgery, the progression of Chronic Kidney Disease (CKD), and the survival chances of patients with Chronic Heart Failure (CHF).

Immunogen

Recombinant protein corresponding to human Lipocalin-2.

Application

Detect Lipocalin using this mouse monoclonal antibody, Anti-Lipocalin-2 Antibody, clone PA348-26.3.5 validated for use in western blotting, IHC & ELISA.
Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Lipoacalin-2 in human bone marrow and stomach tissue lysate.

ELISA Analysis: A representative lot from an independent laboratory detected Lipocalin-2 in indirect ELISA.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blotting in human lung tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Lipocalin-2 in 10 µg of human lung tissue lysate.

Target description

~23 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Wei et al.
Scientific reports, 6, 24378-24378 (2016-04-15)
Extracranial arteriovenous malformations (AVMs) are rare but dangerous congenital lesions arising from direct arterial-venous shunts without intervening capillaries. Progressive infiltration, expansion, and soft tissue destruction lead to bleeding, pain, debilitation and disfigurement. The pathophysiology of AVMs is not well understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service