Skip to Content
Merck
All Photos(1)

Key Documents

MABF2325

Sigma-Aldrich

Anti-Adeno-associated virus 9 Antibody, clone HL2370

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

HL2370
monoclonal

purified by

using protein G

species reactivity

mouse, virus

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
dot blot: suitable

isotype

IgG2aκ

capacity

neutralization

General description

Recombinant Adeno-associated virus serotype 9 capsid protein.

Specificity

Clone HL2370 is a mouse monoclonal antibody that specifically detects AAV9 capsid protein.

Application

Dot Blot Analysis: A representative lot detected Adeno-associated virus 9 in Dot Blot applicaitons (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

Neutralizing Analysis: A representative lot neutralized Adenovirus infection in cells in Neutrailizing applicaitons (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

ELISA Analysis: A representative lot detected Adeno-associated virus 9 in ELISA applicaitons (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

Quality

Evaluated by Dot Blot with AAV8 and AAV9 virus like particles.

Dot Blot Analysis: Various dilutions of AAV8 and AAV9 virus like particles were probed with Adenovirus 9. This antibody specifically detected AAV9, but not AAV8.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Virus (Adeno virus)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service