Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB418PE-I

Sigma-Aldrich

Milli-Mark® Anti-CAR-PE Antibody, clone RmcB

clone RmcB, Milli-Mark®, from mouse

Synonym(s):

46 kD coxsackievirus and adenovirus receptor (CAR) protein, CVB3 binding protein, CVB3-binding protein, Coxsackievirus B-adenovirus receptor, coxsackie virus B receptor, coxsackie virus and adenovirus receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RmcB, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CXADR(1525)

Related Categories

General description

The coxsackievirus and adenovirus receptor (CAR) mediates cell attachment and infection by coxsackie B viruses and by a number of adenoviruses. CAR also mediates homotypic intercellular interactions. In polarized epithelial cells, CAR is closely associated with the tight junction, where it contributes to the barrier to paracellular flow of solutes and macromolecules. CAR′s biological roles are not well defined, but emerging evidence suggests that it may function during embryonic development and in regulating cell proliferation.

Specificity

Antibody recognizes CAR.

Immunogen

Native human coxsackie virus-adenovirus receptor protein

Application

Detect CAR using this Milli-Mark Anti-CAR-PE Antibody, clone RmcB validated for use in FC.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by flow cytometry using HT-29 cells.

Target description

40 kDa Calculated. Uniprot indicates multiple isoforms ranging from 9 to 40 kDa

Physical form

Protein G Purified
Purified mouse monoclonal IgG1 conjugated to PE in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
HT-29 cells

Other Notes

Unconjugated version Cat. No. 05-644 is known to react with Mouse and Rat.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Klichinsky et al.
Nature biotechnology, 38(8), 947-953 (2020-05-04)
Chimeric antigen receptor (CAR) T cell therapy has shown promise in hematologic malignancies, but its application to solid tumors has been challenging1-4. Given the unique effector functions of macrophages and their capacity to penetrate tumors5, we genetically engineered human macrophages
Hongkang Xi et al.
The Journal of experimental medicine, 213(2), 189-207 (2016-01-13)
Age-related macular degeneration (AMD), a leading cause of vision impairment in the ageing population, is characterized by irreversible loss of retinal pigment epithelial (RPE) cells and photoreceptors and can be associated with choroidal neovascularization. Mononuclear phagocytes are often present in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service