Skip to Content
Merck
All Photos(2)

Key Documents

ABE559

Sigma-Aldrich

Anti-Xrcc1 Antibody

serum, from rabbit

Synonym(s):

DNA repair protein XRCC1, X-ray repair cross-complementing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... XRCC1(7515)

General description

DNA repair protein XRCC1 (X-ray repair cross-complementing protein 1) is involved in the efficient repair of DNA single-strand breaks formed by exposure to ionizing radiation and alkylating agents. XRCC1 interacts with DNA ligase III, polymerase beta and poly (ADP-ribose) polymerase to participate in the base excision repair pathway. It may play a role in DNA processing during meiogenesis and recombination in germ cells.

Immunogen

Linear peptide corresponding to mouse Xrcc1.

Application

Anti-Xrcc1 Antibody is a highly specific rabbit polyclonal antibody, that targets XRCC1 & has been tested in western blotting & IP.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Xrcc1 in HepG2 cell lysate.

Western Blotting Analysis: A representative lot from an independent laboratory detected Xrcc1 in HEK293T, HeLa and MEF cell lysates (Lee, Y., et al. (2009). Nature Neurosci. 12:(8):973-980.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory detected Xrcc1 in HEK293T, HeLa and MEF cell lysates (Lee, Y., et al. (2009). Nature Neurosci. 12:(8):973-980.).

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Xrcc1 in 10 µg of Jurkat cell lysate.

Target description

~80 kDa observed. The molecular weight of this protein is calculated at 69 kDa; however it has been observed at ~80 kDa (Lee, Y., et al. (2009). Nature Neurosci. 12:(8):973-980.). An uncharacterized band may be observed at ~51 kDa in some cell lysates.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiaoling Liu et al.
eLife, 10 (2021-06-24)
Targeting DNA repair pathway offers an important therapeutic strategy for Homo sapiens (human) cancers. However, the failure of DNA repair inhibitors to markedly benefit patients necessitates the development of new strategies. Here, we show that exosome component 1 (EXOSC1) promotes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service