Skip to Content
Merck
All Photos(1)

Key Documents

557525

Sigma-Aldrich

Ro-32-0432

A selective cell-permeable protein kinase C inhibitor.

Synonym(s):

Ro-32-0432, 2-{8-[(Dimethylamino)methyl]-6,7,8,9-tetrahydropyrido[1,2-a]indol-3-yl}-3-(1-methyl-1H-indol-3-yl)maleimide, HCl, Bisindolylmaleimide XI, HCl, 3-(8-((dimethylamino)methyl)-6,7,8,9-tetrahydropyrido[1,2- a]indol-10-y1)-4-(1-methyl-1 H-indol-3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28N4O2 · xHCl
CAS Number:
Molecular Weight:
452.55 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C28H28N4O2.ClH/c1-30(2)15-17-10-11-32-20(12-17)13-18-8-9-19(14-24(18)32)25-26(28(34)29-27(25)33)22-16-31(3)23-7-5-4-6-21(22)23;/h4-9,13-14,16-17H,10-12,15H2,1-3H3,(H,29,33,34);1H

InChI key

WMVQGKBSNAIMHI-UHFFFAOYSA-N

General description

A selective cell-permeable protein kinase C inhibitor. Displays about 10-fold greater selectivity for PKCα (IC50 = 9 nM) and 4-fold greater selectivity for PKCβI (IC50 = 28 nM) over PKCε (IC50 = 108 nM). Also prevents T cell-driven chronic inflammatory responses in vivo.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A selective cell-permeable protein kinase C inhibitor. Displays about a 10-fold greater selectivity for PKCα (IC50 = 9 nM) and a 4-fold greater selectivity for PKCβI (IC50 = 28 nM) over PKCε (IC50 = 108 nM). Also prevents T-cell driven chronic inflammatory responses in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKCα
Product does not compete with ATP.
Reversible: no
Target IC50: 9 nM, 28 nM, 108 nM, against PKCα, PKCβI, and PKCε, respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 4 months at -20°C.

Other Notes

Birchall, A.M., et al. 1994. J. Pharmacol. Exp. Ther.268, 922.
Wilkinson, S.E., et al. 1993. Biochem. J.294, 335.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronika Horkova et al.
Nature immunology, 24(1), 174-185 (2022-12-24)
The kinase LCK and CD4/CD8 co-receptors are crucial components of the T cell antigen receptor (TCR) signaling machinery, leading to key T cell fate decisions. Despite decades of research, the roles of CD4-LCK and CD8-LCK interactions in TCR triggering in
Katryna Pampuscenko et al.
Journal of neurochemistry, 154(3), 316-329 (2019-12-14)
Tau is a microtubule-associated protein, found at high levels in neurons, and its aggregation is associated with neurodegeneration. Recently, it was found that tau can be actively secreted from neurons, but the effects of extracellular tau on neuronal viability are unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service