Skip to Content
Merck
All Photos(1)

Key Documents

528315-M

Millipore

PIPPS

A non-metal complexing, zwitterionic buffer with mixed-mode dissociation constants of pKa1 = 3.73 and pKa2 = 7.96 (100 mM, 25°C) PIPPS has no interference at wavelengths longer than 240 nm.

Synonym(s):

PIPPS, Piperazine-N,Nʹ- bis(3-propanesulfonic Acid), Piperazine-N,Nʹ-bis(3-propanesulfonic Acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22N2O6S2
CAS Number:
Molecular Weight:
330.42
UNSPSC Code:
12161703
NACRES:
NA.28

Quality Level

Assay

≥97% dry basis (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (20 °C)

7.96

solubility

water: 1%
0.25 M NaOH: 5%

storage temp.

2-8°C

InChI

1S/C10H22N2O6S2/c13-19(14,15)9-1-3-11-5-7-12(8-6-11)4-2-10-20(16,17)18/h1-10H2,(H,13,14,15)(H,16,17,18)

InChI key

PDLPTSJWDUCMKS-UHFFFAOYSA-N

General description

A non-metal complexing, zwitterionic buffer with mixed-mode dissociation constants of pKa1 = 3.73 and pKa2 = 7.96 (100 mM, 25°C) PIPPS has no interference at wavelengths longer than 240 nm.
A non-metal complexing, zwitterionic buffer with mixed-mode dissociation constants of pKa1 = 3.73 and pKa2 = 7.96 (aqueous solution, 25°C, ionic strength = 100 mM). PIPPS has no interference at wavelengths longer than 240 nm.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yu, Q., et al. 1997. Anal. Biochem. 253, 50.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service