Skip to Content
Merck
All Photos(1)

Key Documents

341607

Sigma-Aldrich

FGF/VEGF Receptor Tyrosine Kinase Inhibitor, PD173074

The FGF/VEGF Receptor Tyrosine Kinase Inhibitor, PD173074 controls the biological activity of FGF/VEGF Receptor Tyrosine Kinase. Primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

FGF/VEGF Receptor Tyrosine Kinase Inhibitor, PD173074, 1- t-Butyl-3-(6-(3,5-dimethoxyphenyl)-2-(4-diethylaminobutylamino)-pyrido[2,3-d]pyrimidin-7-yl)urea, PD173074, VEGFR Tyrosine Kinase Inhibitor XXIII, VEGFR2 Kinase Inhibitor XX, 1-t-Butyl-3-(6-(3,5-dimethoxyphenyl)-2-(4-diethylaminobutylamino)-pyrido[2,3-d]pyrimidin-7-yl)urea, PD173074, VEGFR Tyrosine Kinase Inhibitor XXIII, VEGFR2 Kinase Inhibitor XX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H41N7O3
CAS Number:
Molecular Weight:
523.67
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C28H41N7O3/c1-8-35(9-2)13-11-10-12-29-26-30-18-20-16-23(19-14-21(37-6)17-22(15-19)38-7)25(31-24(20)32-26)33-27(36)34-28(3,4)5/h14-18H,8-13H2,1-7H3,(H3,29,30,31,32,33,34,36)

InChI key

DXCUKNQANPLTEJ-UHFFFAOYSA-N

General description

A cell-permeable pyridopyrimidine compound that acts as a potent, ATP-competitive, and reversible inhibitor of FGF and VEGF receptors (IC50 = 21.5 nM for FGFR1). It inhibits PDGFR and c-Src only at much higher concentration (IC50 = 17.6 µM, 19.8 µM, respectively) and exhibits little effect against EGFR, InsR, MEK, and cPKC even at concentrations as high as 50 µM. Shown to inhibit the autophosphorylation of endogenous FGFR1 (IC50<5 nM) and overexpressed VEGFR2 (IC50<200 nM) in NIH3T3 cells in vitro, and FGF- and VEGF-induced angiogenesis in mice in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Koziczak, M., et al. 2004. Oncogene23, 3501.
Trudel, S., et al. 2004. Blood103, 3521.
Skaper, S.D., et al. 2000. J. Neurochem.75, 1520.
Mohammadi, M., et al. 1998. EMBO J.17, 5896.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jharna R Das et al.
Disease models & mechanisms, 14(7) (2021-07-27)
People of African ancestry living with the human immunodeficiency virus-1 (HIV-1) are at risk of developing HIV-associated nephropathy (HIVAN). Children with HIVAN frequently show high plasma fibroblast growth factor-2 (FGF-2) levels; however, the role of circulating FGF-2 in the pathogenesis
Zhengtian Lyu et al.
Poultry science, 102(1), 102287-102287 (2022-11-29)
Fibroblast growth factor 23 (FGF23), a bone-derived hormone, is involved in the reabsorption of phosphate (P) and the production of vitamin D hormones in the kidney. However, whether and how FGF23 regulates chicken bone metabolism remains largely unknown. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service