Skip to Content
Merck
All Photos(1)

Key Documents

14-445-M

Sigma-Aldrich

Casein Kinase 2α Protein, active, 10 µg

Active, 6His-tagged recombinant full-length Casein Kinase 2α, human, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 48.7 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

General description

6His-tagged recombinant fulllength Casein Kinase 2α, human
Product Source: Expressed in Sf21

Biochem/physiol Actions

Protein Target: CK2α
Target Sub-Family: CMGC

Packaging

Also available in 250μg size as catalog # 14-445M - Call for pricing and availability.

Quality

Casein Kinase 2 Substrate Peptide

Physical form

Ni2+/NTA-agarose

Storage and Stability

1 year at -20°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Casein kinase II beta-subunit inhibits the activity of the catalytic alpha-subunit in the absence of salt.
Tiganis, T, et al.
Biochimica et Biophysica Acta, 1203, 282-289 (1993)
T Tiganis et al.
European journal of biochemistry, 229(3), 703-709 (1995-05-01)
Casein kinase II, cyclin-dependent kinases, and glycogen synthase kinase-3 are members of the protein kinase subfamily with a prominent insert in domain X of their catalytic subunit sequence. The function of the insert sequence in casein kinase II was investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service