Skip to Content
Merck
All Photos(1)

Key Documents

1.09408

Supelco

Buffer solution

(boric acid/potassium chloride/sodium hydroxide)traceable to SRM from NIST and PTB pH 9.00 (25°C) Certipur®

Synonym(s):

Buffer solution pH 9.00 (25°C) (boric acid/potassium chloride/sodium hydroxide)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161705
NACRES:
NA.25

grade

certified reference material

Quality Level

product line

Certipur®

pH

9.0 (25 °C in H2O)

bp

100 °C/1013 hPa

density

1.00 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The boric acid/potassium chloride/sodium hydroxide reference material buffer solution is gravimetrically prepared with high-quality reagents in high-purity water. It is qualified using a combined glass electrode following a 5-point calibration method, in accordance with DIN 19268, against certified primary reference standards as per DIN 19266.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It is used in analytical procedures for the calibration, qualification, and monitoring of pH measuring instruments and as a control for testing other buffer solutions.

Features and Benefits

  • Measured and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to SRMs from NIST and primary certified reference standards from PTB
  • Ready-to-use solution available in a sealed bottle and an innovative Titripac® packaging for a positive environmental impact due to reduced waste generation
  • Long-lasting with a shelf life of 3 years
  • Accompanied with a detailed certificate of analysis (CoA) for an effective audit

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service