1.06578
tri-Sodium phosphate dodecahydrate
for analysis EMSURE® ACS,Reag. Ph Eur
Synonym(s):
tri-Sodium phosphate dodecahydrate, Sodium phosphate tribasic, Trisodium phosphate
About This Item
grade
ACS reagent
Quality Level
Agency
reag. Ph. Eur.
product line
EMSURE®
Assay
98.0-102.0% (alkalimetric)
form
crystals
potency
7400 mg/kg LD50, oral (Rat)
impurities
≤0.001% Total nitrogen (N)
≤0.01% Insoluble matter
pH
12 (20 °C, 10 g/L in H2O)
mp
75 °C (Elimination of water of crystallisation)
solubility
285 g/L
density
1.62 g/cm3 at 20 °C
bulk density
950 kg/m3
anion traces
chloride (Cl-): ≤0.0005%
fluoride (F-): ≤0.0005%
sulfate (SO42-): ≤0.005%
cation traces
Fe: ≤0.001%
excess alkali (as NaOH): ≤2.5%
heavy metals (as Pb): ≤0.001%
storage temp.
2-30°C
SMILES string
[Na+].[Na+].[Na+].[P](=O)([O-])([O-])[O-].O.O.O.O.O.O.O.O.O.O.O.O
InChI
1S/3Na.H3O4P.12H2O/c;;;1-5(2,3)4;;;;;;;;;;;;/h;;;(H3,1,2,3,4);12*1H2/q3*+1;;;;;;;;;;;;;/p-3
InChI key
ASTWEMOBIXQPPV-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
Related Categories
Analysis Note
Insoluble matter: ≤ 0.01 %
excess alkali (as NaOH): ≤ 2.5 %
Chloride (Cl): ≤ 0.0005 %
Fluoride (F): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Corresponds to ACS, Reag. Ph Eur
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea
Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea
Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea
Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service