Skip to Content
Merck
All Photos(1)

Key Documents

1.03872

Millipore

NutriSelect® prime MUELLER HINTON (MH) Agar

acc. FDA-BAM

Synonym(s):

MH Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Quality Level

form

powder

technique(s)

microbiological culture: suitable

pH

7.3±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

nonselective for

General description

NutriSelect® prime MUELLER HINTON (MH) Agar is a special media composition which is designed for the disc diffusion antimicrobial susceptibility testing if common, rapidly growing bacteria from food and animal feed, water, and other samples.
The presence of beef extract and casein hydrolysate provides nitrogen, vitamins, carbon, amino acids, and other nutrients to support the growth of microorganisms. Starch is added to absorb any toxic substances present and agar acts as a solidifying agent.

Application

NutriSelect® prime MUELLER HINTON (MH) Agar is used to test the antibiotic resistance capability of microorganisms in food and animal feed, water, and other samples.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service