Skip to Content
Merck
All Photos(1)

Key Documents

S-098

Supelco

(-)-Scopolamine hydrobromide solution

1.0 mg/mL in acetonitrile: water (9:1) (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO4 · HBr
CAS Number:
Molecular Weight:
384.26
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile: water (9:1) (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[Br].N1([C@@H]2[C@@H]3O[C@@H]3[C@@H]1CC(C2)OC(=O)[C@H](CO)c4ccccc4)C

InChI

1S/C17H21NO4.Br/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10;/h2-6,11-16,19H,7-9H2,1H3;/t11?,12-,13+,14+,15-,16+;/m1./s1

InChI key

XUZPYCBMTSSDHK-AKTDCHNFSA-N

General description

Scopolamine, also known as hyoscine, is a naturally-derived medication used to treat postoperative nausea and vomiting and motion sickness. Produced as a secondary metabolite of plants from the nightshade family, this tropane alkaloid is considered a contaminant in cereal-based foods by the European Commission. This certified solution standard is suitable for use as critical starting materials in calibrators and controls for chromatographic and mass spectrometry testing applications in clinical toxicology, food testing, and forensic analysis.

For the analysis of the phytotoxins. Tropane alkaloid.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service