Skip to Content
Merck
All Photos(1)

Key Documents

W251402

Sigma-Aldrich

Geranyl formate

≥95%, FCC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18O2
CAS Number:
Molecular Weight:
182.26
FEMA Number:
2514
EC Number:
Council of Europe no.:
343
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.076
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Kosher

reg. compliance

FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.46 (lit.)

bp

216 °C (lit.)

density

0.915 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity; rose

SMILES string

C\C(C)=C\CC\C(C)=C\COC=O

InChI

1S/C11H18O2/c1-10(2)5-4-6-11(3)7-8-13-9-12/h5,7,9H,4,6,8H2,1-3H3/b11-7+

InChI key

FQMZVFJYMPNUCT-YRNVUSSQSA-N

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minju Lee et al.
Scientific reports, 8(1), 13945-13945 (2018-09-19)
We report a strategy for the human-like smelling of a rose scent utilizing olfactory receptor nanodisc (ND)-based bioelectronic nose devices. In this strategy, a floating electrode (FE)-based carbon nanotube (CNT) field effect transistor (FET) was functionalized with human olfactory receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service