GF38340200
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3
1m insulated wire, 0.075mm conductor diameter, 0.018mm insulation thickness, ptfe (polytetrafluoroethylene) insulation
Synonym(s):
FE245810
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11171600
NACRES:
NA.23
Recommended Products
description
ptfe (polytetrafluoroethylene) insulation
form
wire (insulated)
manufacturer/tradename
Goodfellow 383-402-00
L
1 m
conductor diam.
0.075 mm
insulation thickness
0.018 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service