Skip to Content
Merck
All Photos(1)

Documents

D104205

Sigma-Aldrich

Dihexyl ketone

97%

Synonym(s):

7-Tridecanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CO(CH2)5CH3
CAS Number:
Molecular Weight:
198.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

264 °C (lit.)

mp

30-32.5 °C (lit.)

density

0.825 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(=O)CCCCCC

InChI

1S/C13H26O/c1-3-5-7-9-11-13(14)12-10-8-6-4-2/h3-12H2,1-2H3

InChI key

ULIAPOFMBCCSPE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Romero-Diaz et al.
Scientific reports, 10(1), 4303-4303 (2020-03-11)
Single substances within complex vertebrate chemical signals could be physiologically or behaviourally active. However, the vast diversity in chemical structure, physical properties and molecular size of semiochemicals makes identifying pheromonally active compounds no easy task. Here, we identified two volatile
Stephanie M Campos et al.
Behavioral ecology : official journal of the International Society for Behavioral Ecology, 31(4), 978-991 (2020-08-09)
Animal signals evolve by striking a balance between the need to convey information through particular habitats and the limitations of what types of signals can most easily be produced and perceived. Here, we present new results from field measures of
Ming Wang et al.
Chemistry, an Asian journal (2018-05-10)
The large polydispersity index of functional pendant polymers has hindered their application in semiconductors. Herein, a novel pendant polymer with perylenediimide (PDI) in the side chains was successfully synthesized through ring-opening metathesis polymerization (ROMP) with a very low polydispersity index.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service