Skip to Content
Merck
All Photos(1)

Key Documents

909416

Sigma-Aldrich

Fluorinated VHL spy molecule 1

Synonym(s):

(2S,4R)-1-(3,3-Dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)-3-(trifluoromethyl)benzyl)pyrrolidine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28F3N3O3S
CAS Number:
Molecular Weight:
483.55
UNSPSC Code:
12352101

ligand

VH032 derivative

form

solid

SMILES string

O=C([C@@H]1C[C@@H](O)CN1C(CC(C)(C)C)=O)NCC2=CC=C(C3=C(C)N=CS3)C(C(F)(F)F)=C2

Application

Fluorinated VHL Spy Molecule 1 can be used for the discovery of new von Hippel-Lindau (VHL) binders. Based on VH032 (cat# 901490), this reporter was designed in the lab of Alessio Ciulli to bind the hydroxyproline (Hyp) recognition site of VHL. When used in competition ligand-observed 19F NMR screening experiments, its displacement indicates binding by another molecule. Additional Fluorinated VHL Spy Molecules are available as listings 909432, 909440 and 909459.
The E3 ligase VHL is of growing interest for targeted protein degradation, and these spy molecules will facilitate the identification of novel VHL ligands and VHL-based degraders.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carles Galdeano et al.
Journal of medicinal chemistry, 57(20), 8657-8663 (2014-08-29)
E3 ubiquitin ligases are attractive targets in the ubiquitin-proteasome system, however, the development of small-molecule ligands has been rewarded with limited success. The von Hippel-Lindau protein (pVHL) is the substrate recognition subunit of the VHL E3 ligase that targets HIF-1α
Guilherme Vieira de Castro et al.
Chemical communications (Cambridge, England), 55(10), 1482-1485 (2019-01-16)
Systematic characterization of a series of fluorinated VHL ligands, varying binding affinity and position of the trifluoromethyl group, qualifies a spy molecule for competitive 19F NMR screening and reveals guiding principles to develop highly sensitive assays with low material consumption.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service