Skip to Content
Merck
All Photos(1)

Key Documents

696951

Sigma-Aldrich

Metronidazole-(ethylene-d4)

98 atom % D, 97% (CP)

Synonym(s):

2-Methyl-5-nitroimidazole-1-ethanol-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D4H5N3O3
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

solid

technique(s)

bio NMR: suitable

mp

159-161 °C (lit.)

mass shift

M+4

SMILES string

[2H]C([2H])(O)C([2H])([2H])n1c(C)ncc1[N+]([O-])=O

InChI

1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3/i2D2,3D2

InChI key

VAOCPAMSLUNLGC-RRVWJQJTSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service