Skip to Content
Merck
All Photos(1)

Key Documents

675156

Sigma-Aldrich

(R)-VANOL

97%

Synonym(s):

(R)-3,3′-Diphenyl-2,2′-bi-1-naphthalol, (R)-3,3′-Diphenyl-2,2′-bi-1-naphthol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H22O2
CAS Number:
Molecular Weight:
438.52
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D +310°, c = 1 in chloroform

mp

200-204 °C

SMILES string

Oc1c(c(cc2ccccc12)-c3ccccc3)-c4c(O)c5ccccc5cc4-c6ccccc6

InChI

1S/C32H22O2/c33-31-25-17-9-7-15-23(25)19-27(21-11-3-1-4-12-21)29(31)30-28(22-13-5-2-6-14-22)20-24-16-8-10-18-26(24)32(30)34/h1-20,33-34H

InChI key

NDTDVKKGYBULHF-UHFFFAOYSA-N

Application

(R)-VANOL is a vaulted biaryl ligand that may be used in the following processes:
  • Transformation of 3-substituted cyclobutanones to enantiopure γ-butyrolactones via asymmetric Baeyer-Villiger Reaction in the presence of an aluminum catalyst.
  • Asymmetric aziridination of N-(4-(Methylsulfonyl)benzylidene)diphenylmethanamine to form (2S,3S)-ethyl 1-benzhydryl-3-(4-(methylsulfonyl)phenyl) aziridine-2-carboxylate.
  • Conjugate addition of terminal alkynes to 2-arylidene-1,3-diketones with high enantioselectivity in the presence of diethyl zinc.
VANOL has proven to be an excellent ligand in catalytic asymmetric Diels-Alder, imine aldol, and aziridination reactions.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vaulted biaryls: efficient ligands for the aluminum-catalyzed asymmetric Baeyer-Villiger reaction.
Bolm C, et al.
Synlett, 2004(09), 1619-1621 (2004)
Blay G, et al.
Chemistry?A European Journal , 18(41), 12966-12969 (2012)
An efficient enantioselective synthesis of florfenicol via asymmetric aziridination.
Wang Z, et al.
Tetrahedron, 67(47), 9199-9203 (2011)
Bao. J. et al.
Journal of the American Chemical Society, 118, 3392-3392 (1996)
Su Yu et al.
Organic letters, 7(3), 367-369 (2005-01-28)
[reaction: see text] In an effort to develop a synthesis of the VAPOL ligand that avoids the use of a chromium carbene complex, a route was examined that involved the annulation of a naphthalene carboxamide via the method of Snieckus.

Related Content

The research areas of interest to the Wulff group include enantioselective catalysis, mechanisms of asymmetric catalysis, macromolecular chemistry, Fischer carbene complexes in organic synthesis and the total synthesis of natural products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service