Skip to Content
Merck
All Photos(1)

Documents

536172

Sigma-Aldrich

Propane

99.5%

Synonym(s):

n-Propane, Propane gas

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH3
CAS Number:
Molecular Weight:
44.10
Beilstein:
1730718
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.5 (vs air)

vapor pressure

190 psi ( 37.7 °C)
8.42 atm ( 21.1 °C)

Assay

99.5%

autoignition temp.

842 °F

expl. lim.

9.5 %

bp

-42.1 °C (lit.)

mp

-188 °C (lit.)

density

0.564 g/mL at 20 °C (lit.)

SMILES string

CCC

InChI

1S/C3H8/c1-3-2/h3H2,1-2H3

InChI key

ATUOYWHBWRKTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propane is an aliphatic hydrocarbon obtained during the oil and natural gas processing. It is generally used as a fuel in automobiles and LPG. It serves as a raw material for the modern chemical industry.

Application

Propane can be used as:
  • A substrate in the study of oxidative dehydrogenation using carbon nanofiber-based catalysts and zeolite catalysts.
  • A component of a binary system along with CO2 to study the solubility of some fat-soluble vitamins.

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-155.2 °F - closed cup

Flash Point(C)

-104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Oxidative dehydrogenation of propane over catalysts based on carbon nanofibers
Sui Z, et al.
Catalysis Today, 106(1-4), 90-94 (2005)
Phase equilibria of the vitamins D2, D3 and K3 in binary systems with CO2 and propane
Knez Z, et al.
Journal of Supercritical Fluids, 20(2), 131-144 (2001)
Oxidative dehydrogenation of propane on zeolite catalysts
Kubacka A, et al.
Catalysis Today, 61(1-4), 343-352 (2000)
The impact of shale gas in the chemical industry
Siirola JJ
AIChE Journal, 60(3), 810-819 (2014)
Walker R Chan et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(14), 5309-5314 (2013-02-27)
The challenging problem of ultra-high-energy-density, high-efficiency, and small-scale portable power generation is addressed here using a distinctive thermophotovoltaic energy conversion mechanism and chip-based system design, which we name the microthermophotovoltaic (μTPV) generator. The approach is predicted to be capable of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service