Skip to Content
Merck
All Photos(1)

Documents

424528

Sigma-Aldrich

4,4′-(Oxydi-2,1-ethanediyl)bismorpholine

97%

Synonym(s):

β,β′-Dimorpholinodiethyl ether, 2,2′-Dimorpholinodiethyl ether, 4,4′-(Oxydiethylene)dimorpholine, Bis(morpholinoethyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24N2O3
CAS Number:
Molecular Weight:
244.33
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

refractive index

n20/D 1.484 (lit.)

bp

309 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

C(CN1CCOCC1)OCCN2CCOCC2

InChI

1S/C12H24N2O3/c1-7-15-8-2-13(1)5-11-17-12-6-14-3-9-16-10-4-14/h1-12H2

InChI key

ZMSQJSMSLXVTKN-UHFFFAOYSA-N

General description

4,4′-(Oxydi-2,1-ethanediyl)bismorpholine (DMDEE) is an amine based catalyst that is also known as dimorpholino-diethyl ether. It can act as a catalyst for blowing reactions and facilitates the process of polymeric curing.

Application

Catalyst for flexible polyester foams, molded foams, and moisture-cured foams and coatings. Good blowing catalyst that does not cause cross-linking.
DMDEE can be used as a property modifier for 3-nitribenzonitrile (3-NDN) which can be further used in matrix assisted ionization vacuum analysis (MAIV). It can also be used as catalyst for formation of polyurethane foams, adhesives and polypropylene glycol (PPG) incorporated fumed silica.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thickening of the Immobilized polymer layer using trace amount of amine and its role in promoting gelation of colloidal nanocomposites.
Zheng Z, et al.
Macromolecules, 48(24), 9015-9023 (2015)
Catalyst-dependent drug loading of LDI-glycerol polyurethane foams leads to differing controlled release profiles.
Sivak WN, et al.
Acta Biomaterialia, 4(5), 1263-1274 (2008)
Structure-property behavior of moisture-cure polyurethane wood adhesives: Influence of hard segment content.
Ren D and Frazier CE
International Journal of Adhesion and Adhesives, 45(2), 118-124 (2013)
Suramya Waidyanatha et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(11), 1341-1351 (2020-06-06)
The specialty amine catalyst 2,2'-dimorpholinodiethyl ether (DMDEE) is a high-production volume chemical used in the production of flexible foam, high-resilient molded foam, and in coatings and adhesives. The disposition and metabolism of [14C]DMDEE (20 or 200 mg/kg) were determined in male
Moisture curing kinetics of isocyanate ended urethane quasi-prepolymers monitored by IR spectroscopy and DSC.
Daniel-da-Silva A, et al.
Journal of Applied Polymer Science, 107(2), 700-709 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service