Skip to Content
Merck
All Photos(1)

Key Documents

394602

Sigma-Aldrich

Iodobenzene-d5

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5I
CAS Number:
Molecular Weight:
209.04
Beilstein:
2502031
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.6161 (lit.)

bp

188 °C (lit.)

mp

-29 °C (lit.)

density

1.868 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(I)c([2H])c1[2H]

InChI

1S/C6H5I/c7-6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D

InChI key

SNHMUERNLJLMHN-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service