Skip to Content
Merck
All Photos(1)

Key Documents

336106

Sigma-Aldrich

2,6-Lutidine

Synonym(s):

2,6-Dimethylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein:
105690
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0 %

Quality Level

form

liquid

refractive index

n20/D 1.497 (lit.)

bp

143-145 °C (lit.)

mp

−6 °C (lit.)

solubility

water: 163.130  g/L at 25 °C

density

0.92 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(C)n1

InChI

1S/C7H9N/c1-6-4-3-5-7(2)8-6/h3-5H,1-2H3

InChI key

OISVCGZHLKNMSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Lutidine, also known as 2,6-dimethylpyridine, is an organic compound that is commonly used as a reagent in various organic reactions, such as the synthesis of heterocycles, nitroalkenes, and alkyl halides. It can also be used as a catalyst in organic synthesis.

Application

2,6-Lutidine can be used as:
  • A base in the synthesis of an aldol adduct from malonic acid hemithioesters and aldehydes catalyzed by Cu(II) salt.
  • An additive in reductive cyclization of epoxygeranyl acetate.
  • A catalyst in combination with CuI for selective synthesis of N-sulfonyl-1,2,3-triazoles.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F

Flash Point(C)

32 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cu (II)-catalyzed enantioselective aldol condensation between malonic acid hemithioesters and aldehydes.
Orlandi S, et al.
Tetrahedron Letters, 45(8), 1747-1749 (2004)
Ti (III)-catalyzed radical cyclization of 6, 7-epoxygeranyl acetate.
Fuse S, et al.
Tetrahedron Letters, 45(9), 1961-1963 (2004)
James R Frost et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(38), 13261-13277 (2015-08-01)
Since their isolation almost 20 years ago, the callipeltosides have been of long standing interest to the synthetic community owing to their unique structural features and inherent biological activity. Herein we present our full research effort that has led to the
Copper-Catalyzed Synthesis of N-Sulfonyl-1, 2, 3-triazoles: Controlling Selectivity.
Yoo EJ, et al.
Angewandte Chemie (International Edition in English), 46(10), 1730-1733 (2007)
Bojana Ginovska et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(44), 15713-15719 (2015-10-24)
We report that 2,6-lutidine⋅trichloroborane (Lut⋅BCl3 ) reacts with H2 in toluene, bromobenzene, dichloromethane, and Lut solvents producing the neutral hydride, Lut⋅BHCl2 . The mechanism was modeled with density functional theory, and energies of stationary states were calculated at the G3(MP2)B3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service