Drug Conjugate Analysis using β-Glucuronidases
Glucuronidation by the human UDP-glucuronosyltransferase (UGT) family of enzymes is important in the metabolic fate of many drugs and other xenobiotics. This biosynthetic reaction also has a role in the conjugation and excretion of endogenous substrates, such as steroids, bilirubin, and bile acids. UGT activity results in the conjugation of glucuronic acid to substrates containing sulfhydryl, hydroxyl, aromatic amino, or carboxylic acid moieties. The glucuronides formed are more polar (water soluble) than the parent organic substrate and are generally excreted through the kidney. Similar conjugation reactions occur with isoforms of sulfotransferases yielding the sulfate conjugate.
Enzymes for Conjugate Hydrolysis
β-Glucuronidase
β-Glucuronidases are routinely used for the enzymatic hydrolysis of glucuronides from urine, plasma, and other biological fluids before analysis by enzyme immunoassay, mass spectrometry, gas chromatography, high performance liquid chromatography, or other means (Figure 1). Typically, between 1 and 20 units of glucuronidase is used per μl of plasma, urine, or bile for the enzymatic hydrolysis of glucuronides present in these samples. The exact amount needed will depend on the specific conditions used and must be determined empirically.
Figure 1. β-Glucuronidase is routinely used to hydrolyze xenobiotic glucuronide metabolites to increase volitility and organic solvent solubility prior to GC analysis.
Molluskan Source β−Glucuronidase
ß-Glucuronidase preparations isolated from mollusks also contain sulfatase activity. For this reason, the sulfatase activity of these preparations is also reported on the certificate of analysis.
The enzyme from patella vulgata is reported to be much more effective in hydrolyzing opioidglucuronides. Whereas the Helix pomatia and E. coli enzymes are reported to be slightly more effective in hydrolyzing steroidglucuronides.
E. coli Source β-GLucuronidase
E. coli derived β-glucuronidase is a ~290 kDa tetrameric protein with an isoelectric point of 4.8. Unlike the enzyme preparations from mollusks that naturally contain β-glucuronidase and sulfatase activities in almost equal amounts, the preparation of β-glucuronidase from E. coli is essentially free of sulfatase activity. The enzyme from E. coli has a high rate of hydrolytic activity and it retains this activity during hydrolysis better than similar enzymes that are more sensitive to changes in the concentration of β-glucuronide conjugates. The enzyme preparation from E. coli has been shown to be useful for determining the presence of androsterone, 17-hydroxycorticosteroids, and estriol in urine. The E. coli enzyme has also been shown to be more active against estrogen conjugates than other sources of the enzyme.
Optimal pH: 6–7
Bovine Liver Source β-GLucuronidase
Bovine ß-glucuronidase is a 290 kD protein with an isoelectric point of 5.1.
Bovine preparations typically contain small amounts of sulfatase activity, usually less than 0.5%.
Optimal pH
β-glucuronidase activity: 4.4
sulfatase activity: 4.4
Sulfatase
Molluskan sources of sulfatase also contain β-glucuronidase activity. Lot-specific activities are reported for both enzymes on the certificate of analysis.
Substrates and Inhibitors | ||
---|---|---|
Name | Description | Cat. No. |
5-Bromo-4-chloro-3-indolyl β-dglucuronide sodium salt | Chromogenic substrate for β-glucuronidase | B8174-5TAB |
5-Bromo-6-chloro-3- indolyl β-d-glucuronide cyclohexylammonium salt | Chromogenic substrate for β-glucuronidase; produces a magenta color in GUS+ bacterial colonies. | B4532-10MG |
d-Glucuronic acid | β-glucuronidase inhibitor | G5269-10MG G5269-10G G5269-25G G5269-50G G5269-100G |
4-Methylumbelliferyl sulfate potassium salt | Fluorescent substrate for sulfatase | M7133-500MG M7133-1G |
4-Nitrocatechol sulfate dipotassium salt | Chromogenic substrate for β-glucuronidase | N7251-100MG N7251-500MG N7251-1G N7251-5G |
4-Nitrophenyl β-d-glucuronide | Chromogenic substrate for β-glucuronidase | N1627-25MG N1627-50MG N1627-100MG N1627-250MG N1627-1G N1627-2G |
Phenolphthalein β-D-glucuronide | Chromogenic substrate for β-glucuronidase | P0501-25MG P0501-100MG P0501-500MG |
Phenolphthalein β-d-glucuronide sodium salt | Chromogenic substrate for β-glucuronidase | P0376-25MG P0376-100MG P0376-250MG P0376-1G |
Network error: Failed to fetch