Merck
HomeSmall Molecules Analysis & QCAssay of Ibuprofen Esters & Impurities
Assay of Ibuprofen Esters & Impurities
Paul Lee, Jenna Milliken
Merck Laramie, WY
INTRODUCTION
Ibuprofen, the active pharmaceutical ingredient in many brand-named products, is one of the most prolific APIs on the market and in our households. This application note describes an analysis of Ibuprofen-related compounds and impurities using an HPLC method similar to the compendial method described by the United States Pharmacopeia and European Pharmacopeia. The described ibuprofen assay method separates ibuprofen from various esters and intermediate impurities on the solid-core Ascentis® Express 90Å C18 column (USP L1) with 0.1% phosphoric acid in both water and acetonitrile as mobile phase. Due to the low absorbance maximum for ibuprofen at 220 nm, a slower gradient maximizes the separation and minimizes baseline drift.
Chemical structures of ibuprofen and ibuprofen-related impurities
Figure 1.Chemical structures of ibuprofen and ibuprofen-related impurities.
EXPERIMENTAL CONDITIONS
Column:
Ascentis® Express C18, 150 x 4.6 mm I.D., 2.7 µm (53829-U)
Mobile Phase:
[A] 0.1% phosphoric acid in water;
[B] 0.1% phosphoric acid in acetonitrile
Gradient:
Time (min)     %A     %B    
0                           48       52      
3                           48       52      
13                        15       85      
16                        15       85      
Flow Rate:
1 mL/min
Pressure:
194 bar (2814 psi)
Temperature:
Column: 30 °C
Autosampler: 12 °C
Detection:
DAD @ 220 nm, 254 nm
Injection:
7 µL
Samples
Diluent:
Acetonitrile and water (50:50)
Sample concentration:
Ibuprofen Standard - 0.4 mg/mL 
Resolution Solution - 0.07 mg/mL (each compound)
Results
Chromatogram of ibuprofen standard 0.4 mg/mL
Figure 2.Chromatogram of ibuprofen standard 0.4 mg/mL.
Chromatogram for Ibuprofen and its related impurities
Figure 3.Chromatogram for ibuprofen and its related impurities.
Peaks
Compound
Retention Time (min)
Relative Retention Time
Resolution to Preceding Peak
USP Tailing
λmax
1
1-(4-Isobutylphenyl)ethanol
5.224
0.92
-
1.07
220 nm                    
2
Ibuprofen
5.679
1.00
3.66
1.10
220 nm
3 | 4
2-Hydroxypropyl 2-(4-Isobutylphenyl)-propanoate)
7.212 | 7.517
1.27 | 1.32
11.70 (2.38)
1.39 | 1.03
220 nm
5
2-(4-Isobutyrylphenyl)propane
9.763
1.72
18.40
0.99
254 nm
6
Ethyl 2-(4-Isobutylphenyl)propanoate
12.696
2.24
24.19
0.99
220 nm
7
4-Isobutylstyrene
13.817
2.43
9.30
1.00
254 nm
8
Isopropyl 2-(4-Isobutylphenyl)propanoate
14.253
2.51
3.66
1.06
220 nm
9
1-Ethyl-4-isobutylbenzene
15.217
2.68
8.11
1.01
220 nm
Calibration and Linearity Data for Ibuprofen and its Related Compounds
Ibuprofen
2-(4-Isobutyrylphenyl)propane
2-Hydroxypropyl 2-(4-Isobutylphenyl)propanoate
Isopropyl 2-(4-Isobutylphenyl)propanoate
Concentration (μg/mL)
Mean Area
Mean Area
Mean Area
Mean Area
0.4
5.8
10.5
4.7
5.0
1.2
17.8
33.1
13.5
16.3
2.0
30.5
55.6
23.0
27.2
2.8
43.0
78.4
32.2
39.6
4.0
60.5
107.8
46.2
55.3
Std Error
0.31
1.28
0.15
0.47
STDEV
0.69
2.87
0.34
1.05
Slope
15.26
27.18
11.58
14.03
LOD (µg/mL)
0.14
0.32
0.09
0.22
LOQ (µg/mL)
0.46
1.05
0.30
0.75
1-Ethyl-4-isobutylbenzene
4-Isobutylstyrene
1-(4-Isobutylphenyl)ethanol
Ethyl 2-(4-Isobutylphenyl)propanoate
Concentration (μg/mL)
Mean Area
Mean Area
Mean Area
Mean Area
0.4
3.7
9.1
6.5
4.7
1.2
11.7
27.7
20.3
16.3
2.0
20.3
47.2
34.4
28.3
2.8
29.4
66.8
47.7
39.7
4.0
41.8
98.5
67.8
56.8
Std Error
0.28
0.91
0.24
0.23
STDEV
0.63
2.03
0.55
0.51
Slope
10.67
24.82
17.03
14.50
LOD (µg/mL)
0.18
0.25
0.10
0.11
LOQ (µg/mL)
0.59
0.82
0.32
0.35
Ibuprofen
2-(4-Isobutyrylphenyl)propane
2-Hydroxypropyl 2-(4-Isobutylphenyl)-propanoate)
Isopropyl 2-(4-Isobutylphenyl)propanoate
Concentration (μg/mL)
Mean Area
Mean Area
Mean Area
Mean Area
0.4
5.8
10.5
4.7
5.0
1.2
17.8
33.1
13.5
16.3
2.0
30.5
55.6
23.0
27.2
2.8
43.0
78.4
32.2
39.6
4.0
60.5
107.8
46.2
55.3
RSQ
0.9998
0.9987
0.9999 | 1.0000
0.9994
Concentration (μg/mL)
Mean Area
Mean Area
Mean Area
Mean Area
100
1516.8
3561.9 (125 µg/mL)
1138.5
1387.9
200
3193.9
7346.3 (250 µg/mL)
2377.4
2841.5
300
4612.4
10740.2 (375 µg/mL)
3567.0
4349.1
400
6202.9
13648.6 (500 µg/mL)
4767.7
5863.3
500
7493.0
-
5828.0
7123.3
RSQ
0.9983
0.9963
0.9993 | 0.9992
0.9990
Combined RSQ
0.9994
0.9988
0.9998 | 0.9998
0.9997
1-Ethyl-4-isobutylbenzene
4-Isobutylstyrene
1-(4-Isobutylphenyl)ethanol
Ethyl 2-(4-Isobutylphenyl)propanoate
Concentration (μg/mL)
Mean Area
Mean Area
Mean Area
Mean Area
0.4
3.9
9.1
6.5
4.7
1.2
11.7
27.7
20.3
16.3
2.0
20.3
47.2
34.4
28.3
2.8
29.4
66.8
47.7
39.7
4.0
41.7
98.5
67.8
56.8
RSQ
0.9994
0.9993
0.9999
0.9999
Concentration (μg/mL)
Mean Area
Mean Area
Mean Area
Mean Area
100
926.6
2011.4
1714.5
1537.4
200
1896.6
4321.2
3526.5
3212.7
300
2940.9
6568.6
5349.3
4748.2
400
3722.4
8838.6
7085.1
6012.3
500
4654.7
11273.2
8683.4
7672.4
RSQ
0.9973
0.9998
0.9993
0.9981
Combined RSQ
0.9992
0.9995
0.9998
0.9993
CONCLUSION
This method demonstrates excellent resolution for Ibuprofen and ibuprofen-impurities. But the stereochemical identification was not included in the scope of the analysis conducted (2-Hydroxypropyl 2-(4-Isobutylphenyl)propanoate comprises two diastereoisomers as synthesized).
The maximal range of linearity is approximately 500 µg/mL, particularly for 2-(4-Isobutyrylphenyl)propane and 4-Isobutylstyrene, which have strong absorption at 254 nm rather than 220 nm. The method enables ibuprofen to be separated from its impurities with excellent peak shape, with limits of quantification of 1 µg/mL or less.
See also Small Molecules Analysis & Quality Control (QC) for more information on other applications or use our chromatogram search.
See more chromatograms on ibuprofen here.