Merck
HomeApplicationsMaterials Science and Engineering Microelectronics & Nanoelectronics
Microelectronics & Nanoelectronics
Illustration of printed circuit board made of electronic components at the micrometer and nanometer scale.
Microelectronics and nanoelectronics are subfields of electronics in which the nominal feature sizes of electronic components are between 100 and 0.1 micrometers in magnitude (microelectronics) or 100 nanometers or smaller (nanoelectronics). The memory storage power of today’s advanced electronic devices has been achieved by significantly increasing the density of microchips. By decreasing the size of field-effect transistors, more components can be fit into integrated circuits, allowing for more powerful and energy-efficient electronic devices with reduced weights and power consumption.
Something went wrong, please try again.
Featured Categories
Solution deposition and vapor deposition are two synthesis routes used for formation of advanced, precision thin films and coatings.
Solution & Vapor Deposition Precursors
Our high-quality solution deposition and vapor deposition precursors are optimal for...
View Products
Self-Assembly & Contact Printing
Self-Assembly & Contact Printing
We offer a comprehensive portfolio of self-assembly and nanoimprinting materials for effective target surface modification and precise transferal of micropatterns and nanopatterns to enable high-performance micro- and nanoelectronics.
View Products
OFET & OPV Materials
OFET & OPV Materials
We provide a comprehensive portfolio of OFET and OPV materials and inks including organic semiconductors, organic conductors, dielectric materials and inorganic transport materials, as well as pre-patterned substrates and device components.
View Products
Electronic Chemicals & Etchants
Electronic Chemicals & Etchants
We offer a broad range of electronic chemicals and etchants used in electronic device fabrication, such as electroplating, lithography, etching, doping and packaging.
View Products
According to Moore’s Law, the number of transistors that can be put on a single chip will double every two years. Since this was projected in 1965, semiconductor fabrication technology sustained this rate of advancement and revolutionized the industry. However, pace of dimension reduction is slowing, and the key challenge in fabricating electronic components in the sub-micrometer range is the design of the transistor gate, which controls the current flow in the channel. The smaller electronic components are, the more challenging they become to manufacture. Physical and quantum effects alter materials’ properties from a macroscale to a nanoscale, influencing inter-atomic interactions and quantum mechanical properties.
The arrival of innovative materials, carbon nanotubes, boron nitride nanotubes, quantum dots, and graphene additives, have advanced the minimization of nanotechnology and microtechnology. These and other new materials can be shaped and manipulated with extraordinary precision at the tiniest of scales. Novel technologies enable the deposition and layering of electronic materials with precise thickness, even down to the atomic level. Thin-film semiconductor device fabrication technology uses conducting, semiconducting, and insulating materials to deliver advanced capabilities at high volumes and very low cost. Modern manufacturing methods for nanoelectronics include patterning (lithography), etching, thin film deposition, and doping techniques.
Emerging research fields focus on new approaches in nanotechnology and quantum mechanical effects. Molecular electronics uses single molecules as electronic components to establish electrical contact with bulk-sized electrodes. Spintronics, or spin-transport electronics, manipulates electrons’ spin property with magnetic and electric fields, resulting in a spin-polarized current that provides higher data transfer speeds and greater storage capacity, memory density and processing power than is possible with electric charge alone.
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents