Potassium Organotrifluoroborates
Organotrifluoroborates: Versatile and Stable Boronic Acid Surrogates
Prof. Gary Molander has extensively developed the chemistry of organotrifluoroborates. These bench stable boronic acid surrogates are useful for Suzuki-Miyaura cross-coupling reactions and have also been used for a variety of other C-C bond forming reactions. Importantly, these reagents are compatible with a wide range of functional groups and are stable to many commonly used and harsh reaction conditions.
Benefits of Organtrifluoroborates:
- Bench stable reagents that can be stored indefinitely
- Can be effectively coupled with a wide range of organic electrophiles
- Exist as tetracoordinated monomeric species
- Are less prone to protodeboronation than their boronic acid or boronate ester counterparts
![Suzuki-Miyaura Cross-Coupling with Organotrifluoroborates Suzuki-Miyaura Cross-Coupling with Organotrifluoroborates](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/chemistry-and-synthesis/reaction-design-and-optimization/suzuki-miyaura-cross-coupling-organotrifluoroborates.gif)
Suzuki-Miyaura Cross-Coupling with Organotrifluoroborates
![The Versatility of Organotrifluoroborates Organotrifluoroborates from us](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/chemistry-and-synthesis/reaction-design-and-optimization/veratility-organotrifluoroborates.gif)
Organotrifluoroborates from us
For a complete list of organotrifluoroborates.
Network error: Failed to fetch