Merck
HomeOrganic Reaction ToolboxVisible-Light-Mediated Reactions under Mild Conditions
Visible-Light-Mediated Reactions under Mild Conditions
Anna Lee
Department of Chemistry, Jeonbuk National University
567, Baekje-daero, Deokjin-gu Jeonju 54896, Republic of Korea
Abstract
Visible-light-mediated reactions have been incorporated into various organic synthesis schemes. Recently, owing to the important focus on environmental issues, the development of an environmentally friendly and mild synthetic methodology has become urgently needed. In this review, we summarize our recent approaches to developing the green mode of visible-light-mediated reactions that are available under mild reaction conditions without the use of toxic transition-metal-based catalysts or harsh reaction conditions. Relevant applications in the synthesis of valuable small molecules, including bioactive compounds, have also been highlighted.
Read more about
Introduction
The use of visible light to effect chemical transformations has been a long-term challenge for chemists. As an example, in 1912, Giacomo Ciamician reported “The Photochemistry of the Future” in Science.1 Afterward, Kellogg provided a basis for modern photoredox catalysis by reporting a reductive amination reaction in 1978.2 Interestingly, however, not many studies were reported until the 2010s. Recently, visible-light-mediated photoredox catalysis has attracted a great deal of attention in organic synthesis.3 In these types of reactions, visible light is converted into chemical energy by engaging in single-electron transfer (SET) with substrates to generate reactive intermediates. Based on the unique reaction mode of visible-light-mediated transformations, a wide range of reactions have been developed over the past several years.
In recent years, green activation modes have been explored in this field owing to the growing interest in the development of environmentally benign synthetic methods. The most common and general approach to green photoredox catalysis is the use of organic photocatalysts. Various organophotocatalysts, including newly developed catalysts and traditional organic dyes, such as rose bengal, eosin Y, and 4CzIPN (2,4,5,6-tetrakis(9H-carbazol-9-yl)-isophthalonitrile) (Figure 1), have been employed in numerous reactions.3b,4
Chemical structures of commonly used organic photocatalysts, A) 4CzIPN B) MeS-Acr-Me+ C) Riboflavin D) Rose Bengal E) Eosin Y F) Rhodamine B G)TPP+
Figure 1.Examples of common organic photocatalysts, (A) 4CzIPN (B) MeS-Acr-Me+
(C) Riboflavin (D) Rose Bengal (E) Eosin Y (F) Rhodamine B (G)TPP+.
Recently, many research groups have focused on the advantages of organic photocatalysts because their structures can be modified by a rational approach to realize unprecedented reactivity or improve the catalytic activity. As an alternative green approach, catalyst-free reactions have been developed. These latter types of reaction modes include the use of photoactive starting compounds or in situ excitation of the substrates under visible-light irradiation. The substrates thus activated form electron donor−acceptor (EDA) complexes, obviating the need for photoredox catalysts.5 This strategy relies on associating electron-donor with electron-acceptor substrates, such as Lewis bases and acids, respectively. The EDA complex can absorb visible light, undergoing an excitation process, which triggers a single-electron transfer (SET) that can generate radical intermediates. In addition to the above-mentioned approaches, green synthetic methods using natural substances such as molecular oxygen have also been studied. Our group has focused on the development of novel, visible-light-mediated reactions under mild reaction conditions. Herein, we summarize our approaches for realizing the green mode of visible-light-mediated reactions (Figure 2).
An illustration demonstrating two green approaches (a) catalyst free reactions (b) organo-photocatalysis, for the development of mild, visible-light-mediated reactions.
Figure 2. Our green approaches for the development of mild, visible-light-mediated reactions, (a) Catalyst Free Reactions (b) Organo-photocatalysis
Conclusions and Outlook
The development of green synthetic methods is an important goal in the field of visible-light-mediated reactions. In this short review, we introduced our approaches for carrying out valuable organic transformations under mild reaction conditions such as visible-light-mediated aerobic oxidations using molecular oxygen. Additionally, we expanded the scope of singlet-oxygen-mediated reactions by developing oxidative C–S bond cleavage reactions. Moreover, using EDA complex-based synthetic strategies, we developed catalyst-free reactions through the intermediacy of binary or ternary EDA complexes. Various mechanistic studies have been conducted to understand the reaction mechanisms. However, further studies are required to directly analyze EDA complexes. In this review, applications of these approaches to the synthesis of valuable small molecules were also highlighted. Further studies are in progress in our laboratory to develop novel visible-light-mediated syntheses based on green approaches. Future directions would include the structural modification of organic photocatalysts and the development of novel visible-light-active starting compounds to expand the range of available reactions in this field.
Acknowledgment
Anna Lee thanks her co-workers for their contributions to the development of the reactions described in this article. This research was supported by a National Research Foundation of Korea (NRF) grant funded by the Korean government (MIST) (2023R1A2C1005301 and 2022R1F1A1064110).
References
1.
Ciamician G. 1912. The Photochemistry of the Future. Science. 36(926):385-394. https://doi.org/10.1126/science.36.926.385
2.
Hedstrand DM, Kruizinga WH, Kellogg RM. 1978. Light induced and dye accelerated reductions of phenacyl onium salts by 1,4-dihydropyridines. Tetrahedron Lett. 19(14):1255-1258. https://doi.org/10.1016/s0040-4039(01)94515-0
3.
For selected reviews on photoredox catalysis, see: (a) Prier CK, Rankic DA, MacMillan DWC. 2013. Visible Light Photoredox Catalysis with Transition Metal Complexes: Applications in Organic Synthesis. Chem. Rev. 113(7):5322-5363. https://doi.org/10.1021/cr300503r (b) Romero NA, Nicewicz DA. 2016. Organic Photoredox Catalysis. Chem. Rev. 116(17):10075-10166. https://doi.org/10.1021/acs.chemrev.6b00057 (c) Shaw MH, Twilton J, MacMillan DWC. 2016. Photoredox Catalysis in Organic Chemistry. J. Org. Chem. 81(16):6898-6926. https://doi.org/10.1021/acs.joc.6b01449 (d) Marzo L, Pagire SK, Reiser O, König B. 2018. Visible-Light Photocatalysis: Does It Make a Difference in Organic Synthesis?. Angew. Chem.Int. Ed. 57(32):10034-10072. https://doi.org/10.1002/anie.201709766 (e) Mateus-Ruiz JB, Cordero-Vargas A. 2020. Visible-Light-Mediated Photoredox Reactions in the Total Synthesis of Natural Products. Synthesis. 52(21):3111-3128. https://doi.org/10.1055/s-0040-1707225 (f) Reed NL, Yoon TP. 2021. Oxidase reactions in photoredox catalysis. Chem. Soc. Rev. 50(5):2954-2967. https://doi.org/10.1039/d0cs00797h (g) Yu XY, Chen JR, Xiao WJ. 2021. Visible Light-Driven Radical-Mediated C–C Bond Cleavage/Functionalization in Organic Synthesis. Chem. Rev. 121(1):506-561. https://doi.org/10.1021/acs.chemrev.0c00030 (h) Pitre SP, Overman LE. 2022. Strategic Use of Visible-Light Photoredox Catalysis in Natural Product Synthesis. Chem. Rev. 122(2):1717-1751. https://doi.org/10.1021/acs.chemrev.1c00247
4.
Vega‐Peñaloza A, Mateos J, Companyó X, Escudero‐Casao M, Dell'Amico L. 2021. A Rational Approach to Organo‐Photocatalysis: Novel Designs and Structure‐Property Relationships. Angew. Chem., Int. Ed. 60(3):1082-1097. https://doi.org/10.1002/anie.202006416
5.
(a) Mulliken RS. 1950. Structures of Complexes Formed by Halogen Molecules with Aromatic and with Oxygenated Solvents. J. Am. Chem. Soc. 72(1):600-608. https://doi.org/10.1021/ja01157a151 (b) Miller JS, Epstein AJ, Reiff WM. 1988. Ferromagnetic molecular charge-transfer complexes. Chem. Rev. 88(1):201-220. https://doi.org/10.1021/cr00083a010 (c) Rosokha SV, Kochi JK. 2008. Fresh Look at Electron-Transfer Mechanisms via the Donor/Acceptor Bindings in the Critical Encounter Complex. Acc. Chem. Res. 41(5):641-653. https://doi.org/10.1021/ar700256a (d) Miyasaka H. 2013. Control of Charge Transfer in Donor/Acceptor Metal–Organic Frameworks. Acc. Chem. Res. 2013, 46(2):248-257. https://doi.org/10.1021/ar300102t (e) Lima CGS, de M Lima T, Duarte M, Jurberg ID, Paixão MW. 2016. Organic Synthesis Enabled by Light-Irradiation of EDA Complexes: Theoretical Background and Synthetic Applications. ACS Catal. 6(3):1389-1407. https://doi.org/10.1021/acscatal.5b02386 (f) De A Bartolomeu A, Silva RC, Brocksom TJ, Noël T, de Oliveira KT. 2019. Photoarylation of Pyridines Using Aryldiazonium Salts and Visible Light: An EDA Approach. J. Org. Chem. 84(16):10459-10471. https://doi.org/10.1021/acs.joc.9b01879
6.
(a) Cao Q, Dornan LM, Rogan L, Hughes NL, Muldoon MJ. 2014. Aerobic oxidation catalysis with stable radicals. Chem. Commun. 50(35):4524-4543. https://doi.org/10.1039/c3cc47081d (b) Kananovich, D.; Elek GZ, Lopp M, Borovkov V. 2021. Aerobic Oxidations in Asymmetric Synthesis: Catalytic Strategies and Recent Developments. Front. Chem. 9:614944. https://doi.org/10.3389/fchem.2021.614944 (c) Liu J, Guðmundsson A, Bäckvall JE. 2021. Efficient Aerobic Oxidation of Organic Molecules by Multistep Electron Transfer. Angew. Chem., Int. Ed. 60(29):15686-15704. https://doi.org/10.1002/adsc.201800730
7.
Jiang J, Luo R, Zhou X, Chen Y, Ji H. 2018. Photocatalytic Properties and Mechanistic Insights into Visible Light‐Promoted Aerobic Oxidation of Sulfides to Sulfoxides via Tin Porphyrin‐Based Porous Aromatic Frameworks. Adv. Synth. Catal. 360(22):4402-4411. https://doi.org/10.1002/adsc.201800730
8.
Kim DH, Lee J, Lee A. 2018. Visible-Light-Driven Silver-Catalyzed One-Pot Approach: A Selective Synthesis of Diaryl Sulfoxides and Diaryl Sulfones. Org. Lett. 20(3):764-767. https://doi.org/10.1021/acs.orglett.7b03901
9.
(a) Emmett EJ, Hayter BR, Willis MC.2013. Palladium-Catalyzed Three-Component Diaryl Sulfone Synthesis Exploiting the Sulfur Dioxide Surrogate DABSO. Angew. Chem., Int. Ed. 125(48):12679-12683. https://doi.org/10.1002/anie.201305369 (b) Margraf N, Manolikakes G. 2015. One-Pot Synthesis of Aryl Sulfones from Organometallic Reagents and Iodonium Salts. J. Org. Chem. 80(5):2582-2600. https://doi.org/10.1021/jo5027518
10.
For selected reviews on silver catalysis, see: (a) Dénès F, Pichowicz M, Povie G, Renaud P. 2014. Thiyl Radicals in Organic Synthesis. Chem. Rev. 114(5):2587-2693. https://doi.org/10.1021/cr400441m (b) Fang G, Cong X, Zanoni G, Liu Q, Bi X. 2017. Silver-Based Radical Reactions: Development and Insights. Adv. Synth. Catal. 359(9):1422-1502. https://doi.org/10.1002/adsc.201601179
11.
(a) DeRosa MC, Crutchley RJ. 2002. Photosensitized singlet oxygen and its applications. Coord. Chem. Rev. 233–234:351-371. https://doi.org/10.1016/S0010-8545(02)00034-6 (b) Lemp E, Valencia C, Zanocco AL. 2004. Solvent effects on reactions of singlet molecular oxygen, O2(1Δg), with antimalarial drugs. J. Photochem. Photobiol. A. 168(1-2):91-96. https://doi.org/10.1016/j.jphotochem.2004.05.016
12.
Narayanam JMR, Stephenson CRJ. 2011. Visible light photoredox catalysis: applications in organic synthesis. Chem. Soc. Rev. 40(1):102-113. https://doi.org/10.1039/b913880n
13.
Aganda KCC, Hong B, Lee A. 2019. Aerobic α‐Oxidation of N‐Substituted Tetrahydroisoquinolines to Dihydroisoquinolones via Organo‐photocatalysis. Adv. Synth. Catal. 361(5):1124-1129. https://doi.org/10.1002/adsc.201801301
14.
Bancirova M. 2011. Sodium azide as a specific quencher of singlet oxygen during chemiluminescent detection by luminol and Cypridina luciferin analogues. Luminescence. 26(6):685-688. https://doi.org/10.1002/bio.1296
15.
(a) Möbius K. 2000. Primary processes in photosynthesis: what do we learn from high-field EPR spectroscopy?. Chem. Soc. Rev. 29(2):129-139. https://doi.org/10.1039/a706426h (b) Rieger AL, Rieger PH. 2004. Chemical Insights from EPR Spectra of Organometallic Radicals and Radical Ions. Organometallics. 23(2):154-162. https://doi.org/10.1021/om030565e (c) Roessler MM, Salvadori E. 2018. Principles and applications of EPR spectroscopy in the chemical sciences. Chem. Soc. Rev. 47(8):2534-2553. https://doi.org/10.1039/c6cs00565a
16.
Wasserman HH, Ives JL. 1981. Singlet oxygen in organic synthesis. Tetrahedron Lett. 37(10):1825-1852. https://doi.org/10.1016/s0040-4020(01)97932-3
17.
Corey E, Ouannés C. 1976. Intermédiaire dans l'oxydation de benzylalcoyl sulfures par l'oxygène singulet. Tetrahedron Lett. 17(47):4263-4266. https://doi.org/10.1016/0040-4039(76)80090-1
18.
(a) Pasto DJ, Cottard F, Jumelle L. 1994. Photooxidation of Alkyl 4-Nitrophenyl Sulfides and Sulfoxides. Observation of Oxidative C-S Bond Cleavage and Rearrangement Reactions. J. Am. Chem. Soc. 116(20):8978-8984. https://doi.org/10.1021/ja00099a015 (b) Jensen F, Greer A, Clennan EL. 1998. Reaction of Organic Sulfides with Singlet Oxygen. A Revised Mechanism. J. Am. Chem. Soc. 120(18):4439-4449. https://doi.org/10.1021/ja973782d (c) Toutchkine A, Clennan EL. 2000. A New Mechanism for Oxidative C−S Bond Cleavage during Reactions of Singlet Oxygen with Organic Sulfides:  Electronically Dictated Reaction Selectivity in the Persulfoxide Intermediate. J. Am. Chem. Soc. 122(8):1834-1835. https://doi.org/10.1021/ja993867 (d) Clennan EL. 2001. Persulfoxide: Key Intermediate in Reactions of Singlet Oxygen with Sulfides. Acc. Chem. Res. 34(11):875-884. https://doi.org/10.1021/ar0100879 (e) Toutchkine A, Aebisher D, Clennan EL. 2001. Substituent-Dictated Partitioning of Intermediates on the Sulfide Singlet Oxygen Reaction Surface. A New Mechanism for Oxidative C−S Bond Cleavage in α-Hydroperoxy Sulfides. J. Am. Chem. Soc. 123(21):4966-4973. https://doi.org/10.1021/ja004188y (f) Bonesi SM, Fagnoni M, Monti S, Albini A. 2006. Reaction of singlet oxygen with some benzylic sulfides. Tetrahedron Lett. 62(46):10716-10723. https://doi.org/10.1016/j.tet.2006.07.110 (g) Baciocchi E, Del Giacco T, Lanzalunga O, Lapi A. 2007. Singlet Oxygen Promoted Carbon−Heteroatom Bond Cleavage in Dibenzyl Sulfides and Tertiary Dibenzylamines. Structural Effects and the Role of Exciplexes. J. Org. Chem. 72(25):9582-9589. https://doi.org/10.1021/jo701641b (h) Baciocchi E, Del Giacco T, Lanzalunga O, Mencarelli P, Procacci B. 2008. Photosensitized Oxidation of Alkyl Phenyl Sulfoxides. C−S Bond Cleavage in Alkyl Phenyl Sulfoxide Radical Cations. J. Org. Chem. 73(15):5675-5682. https://doi.org/10.1021/jo801088n
19.
(a) Vosburgh WC, Cogswell SA. 1943. Complex Ions. VIII. Pyridine—Silver Ions. J. Am. Chem. Soc. 65(12):2412-2413. https://doi.org/10.1021/ja01252a049 (b) Po HN. 1976. Heterocyclic and macrocyclic amine complexes of silver(II) and silver(III). Coord. Chem. Rev. 20(2):171-195. https://doi.org/10.1016/S0010-8545(00)80324-0
20.
Hong B, Aganda KCC, Lee A. 2020. Oxidative C–S Bond Cleavage of Benzyl Thiols Enabled by Visible-Light-Mediated Silver(II) Complexes. Org. Lett. 22(11):4395-4399. https://doi.org/10.1021/acs.orglett.0c01399
21.
Hong B, Lee A. 2022. Visible-light-mediated oxidative C–S bond cleavage of benzyl thiols through in situ activation strategy. Org. Biomol. Chem. 20(30):5938-5942. https://doi.org/10.1039/d2ob00089j
22.
Ritu, Sharma C, Kumar S, Jain N. 2020. Singlet oxygen mediated dual C–C and C–N bond cleavage in visible light. Org. Biomol. Chem. 18(15):2921-2928. https://doi.org/10.1039/d0ob00563k
23.
Lee J, Hong B, Lee A. 2019. Visible-Light-Promoted, Catalyst-Free Gomberg–Bachmann Reaction: Synthesis of Biaryls. J. Org. Chem. 84(14):9297-9306. https://doi.org/10.1021/acs.joc.9b00557
24.
Gomberg M, Bachmann WE. 1924. THE SYNTHESIS OF BIARYL COMPOUNDS BY MEANS OF THE DIAZO REACTION. J. Am. Chem. Soc. 46(10):2339-2343. https://doi.org/10.1021/ja01675a026
25.
(a) MacCarthy P. 1978. Simplified experimental route for obtaining Job's curves. Anal. Chem. 50(14):2165. https://doi.org/10.1021/ac50036a059 (b) Renny JS, Tomasevich LL, Tallmadge EH, Collum DB. 2013. Method of Continuous Variations: Applications of Job Plots to the Study of Molecular Associations in Organometallic Chemistry. Angew. Chem., Int. Ed. 52(46):11998-12013. https://doi.org/10.1002/anie.201304157
26.
Benesi HA, Hildebrand JH. 1949. A Spectrophotometric Investigation of the Interaction of Iodine with Aromatic Hydrocarbons. J. Am. Chem. Soc. 71(8):2703-2707. https://doi.org/10.1021/ja01176a030
27.
(a) Masnovi JM, Kochi JK, Hilinski EF, Rentzepis PM. 1985. Observation of ternary electron donor-acceptor complexes of anthracene by picosecond spectroscopy. Molecular structure of the anthracenophane-tetracyanoethylene complex. J. Phys. Chem. 89(25):5387-5395. https://doi.org/10.1021/j100271a015 (b) Woźniak Ł, Murphy JJ, Melchiorre P. 2015. Photo-organocatalytic Enantioselective Perfluoroalkylation of β-Ketoesters. J. Am. Chem. Soc. 137(17):5678-5681. https://doi.org/10.1021/jacs.5b03243 (c) Sandfort F, Strieth- Kalthoff F, Klauck FJR, James MJ, Glorius F. 2018. Deaminative Borylation of Aliphatic Amines Enabled by Visible Light Excitation of an Electron Donor–Acceptor Complex. Chem.—Eur. J. 24(65):17210-17214. https://doi.org/10.1002/chem.201804246 (d) Wu J, Grant PS, Li X, Noble A, Aggarwal VK. 2019. Catalyst-Free Deaminative Functionalizations of Primary Amines by Photoinduced Single-Electron Transfer. Angew. Chem., Int. Ed. 58(17):5697-5701. https://doi.org/10.1002/anie.201814452 (e) Kim JY, Lee YS, Ryu DH. 2021. Ternary Electron Donor–Acceptor Complex Enabled Enantioselective Radical Additions to α, β-Unsaturated Carbonyl Compounds. ACS Catalysis. 11(24):14811-14818. https://doi.org/10.1021/acscatal.1c04835
28.
Aganda KCC, Kim J, Lee A. 2019. Visible-light-mediated direct C3-arylation of 2H-indazoles enabled by an electron-donor–acceptor complex. Org. Biomol. Chem. 17(45):9698-9702. https://doi.org/10.1039/c9ob02074h
29.
Aganda KCC, Hong B, Lee A. 2021. Visible‐Light‐Promoted Switchable Synthesis of C‐3‐Functionalized Quinoxalin‐2(1H)‐ones. Adv. Synth. Catal. 363(5):1443-1448. https://doi.org/10.1002/adsc.202001396
30.
Aganda KCC, Lee A. 2021. Synthesis of Selenaheterocycles via Visible‐Light‐Mediated Radical Cyclization. Adv. Synth. Catal. 363(22):5149-5154. https://doi.org/10.1002/adsc.202100969