Merck
HomeStem Cell CultureMesenchymal Stem Cell Culture
Mesenchymal Stem Cell FBS, Media, and Culture
Mesenchymal stem cell overview
Figure 1. Mesenchymal stem cell overview.Trilineage differentiation of multipotent human mesenchymal stem cells (MSCs) includes differentiation into osteoblasts, adipocytes and chondrocytes.
Human mesenchymal stem cells (MSCs) are multipotent adult stem cells. MSCs have the capacity for multi-lineage differentiation, giving rise to a variety of mesenchymal phenotypes such as osteoblasts, adipocytes, and chondrocytes. Mesenchymal stem cells usually reside in the bone marrow stroma, but have also been found in adipose, liver, spleen, peripheral blood, umbilical cord blood, and other mesenchymal tissues. MSCs have the capacity for self-renewal over long periods of time and the ability to differentiate into specialized cells. This has led to an increased interest in understanding the biology and therapeutic potential of MSC cultures by researchers.
We offer a broad range of tools and technologies, such as optimized expansion and differentiation media, to culture mesenchymal stem cells. Our collection also includes low passage MSC lines from various tissues, specialty FBS, and a broad range of MSC related antibodies and cellular dyes.
Mesenchymal Stem Cell Lines
Our extensive collection of mesenchymal stem cell lines provides primary cell lines from multiple tissue sources. These human tissue sources include bone marrow, adipose, and umbilical cord tissue-sourced MSCs. We also offer MSC lines from human embryonic stem cells (ES) and rat bone marrow tissue.
Ordering Info
Cell TypeBrandFormatPart Number
Human Bone-Marrow MSCsPromoCell*5X105 Cryopreserved CellsC-12974
 Sigma1X106 Cryopreserved CellsSCC034
 PromoCell*5X105 Live Proliferating CellsC-12975
 PromoCell*1X106 Cell PelletC-14090
Human Adipose MSCsPromoCell*5X105 Cryopreserved CellsC-12977
 Sigma1X106 Cryopreserved CellsSCC038
 PromoCell*5X105 Live Proliferating CellsC-12978
 PromoCell*1X106 Cell PelletC-14092
Human Umbilical Cord MSCsPromoCell*5X105 Cryopreserved CellsC-12971
 PromoCell*5X105 Live Proliferating CellsC-12972
 PromoCell*1X106 Cell PelletC-14091
Human ES Cell Derived MSCsSigma1X106 Cryopreserved CellsSCC036
Rat Bone-Marrow MSCsSigma1X106 Cryopreserved CellsSCR027
Human mesenchymal stem cell markers
Figure 2. Human MSC markers.Human MSCs express H-CAM (CD44) (A), THY-1 (CD90) (B), and STRO-1 (C). Nuclei were visualized with DAPI (blue). Expression of hematopoietic stem cell markers CD19 and CD14 and endothelial marker CD146 were not observed (data not shown).
Mesenchymal Stem Cell Expansion Media
MSC Expansion Media
Our collection of MSC qualified media includes a variety of high performing complete media for expansion of mesenchymal stem cells. From low-serum to xeno-free media options, we offer a solution to every type of MSC researcher. To learn how to expand MSCs, click here.
Ordering Info
Serum ContainingDefined/Xeno-Free
Low FBS (2%) cell culture media for human mesenchymal stem cells.
Mesenchymal Stem Cell Growth Medium 2 (PromoCell)
Mesenchymal Stem Cell Expansion Medium (Sigma)
Complete xeno-free and serum-free media that is free of animal-derived components.
Mesenchymal Stem Cell Growth Medium DXF (PromoCell)
Human Mesenchymal-XF Expansion Medium (Sigma)
MESENCHYMAL STEM CELL DIFFERENTIATION MEDIA
Our validated MSC differentiation media portfolio is optimized for differentiation into a variety of mesenchymal lineages. The collection includes media designed for the directed differentiation of MSCs into osteogenic (bone), adipogenic (fat), or chondrogenic (cartilage) lineages. For more information on how to differentiate MSCs, click here.
PLTMax® Human Platelet Lysate
A Superior Alternative to FBS for Human MSC Cultures
With the increasing interest and utilization of human cellular therapy, PLTMax® Human Platelet Lysate meets the need for a research grade cell culture supplement. The FBS alternative is human derived, serum-free, and cost effective for human mesenchymal stem cell cultures.
Application Note: PLTMax® Human Platelet Lysate (hPL): A Superior Serum-Free Media Supplement for Robust Mesenchymal Stem Cell Expansion
Features and Benefits:
  • Increased cell growth kinetics vs. FBS supplemented media
  • Cost effective alternative to FBS-supplemented serum free media
  • Manufactured to reduce lot-to-lot variation
  • Human derived material applicable for translational research applications
  • Each of the donor units are tested for infectious diseases such as HIV-1, HIV-2, HCV, and HBsAg, among others. 
PLTMax Human Platelet Lysate
PLTMax supports better growth of hMSCs than DMEM + FBS
Figure 3. PLTMax supports better growth of hMSCs than DMEM + FBSA: Growth of hMSCs cultured in αMEM supplemented with 5 % HPL or 10 % FBS as compared to DMEM + 10 % FBS over 4 passages in T150 flasks, or B: in duplicate 125 mL spinner flasks f or 7 days, demonstrating enhanced growth in αMEM + HPL
Product No.Description
SCM141PLTMax Human Platelet Lysate, 100ml
SCM142PLTMax Human Platelet Lysate, 500ml
SCM151PLTGold Human Platelet Lysate (Heparin-Free), 100ml
SCM152PLTGold Human Platelet Lysate (Heparin-Free), 500ml
MSC Qualified Fetal Bovine Serum
Nutrient-rich FBS is required for some stem cell applications. As a widely used cell culture media supplement, there are many different FBS products to choose from. MSC-qualified FBS (ES-020-B) has been screened using a rigorous clonal growth assay to ensure support for multipotent human MSC cultures over multiple passages.
Features and Benefits:
  • Prescreened to validate for use in human MSC growth
  • Lot reservation upon request
Mesenchymal stem cell growth using various FBS sources.
Figure 4.Mesenchymal stem cell growth using various FBS sources.
Human adipose-derived mesenchymal stem cells (SCC038) were cultured for three passages in media containing 10% FBS from various suppliers. Population doubling times were calculated at each passage. Media supplemented with ES-020-B produced consistent cell growth characteristics with low doubling times over multiple passages compared with doubling times observed using media containing FBS from other vendors. (Figure 4.)
Sorry, an unexpected error has occurred
Network error: Failed to fetch
OsteoMAX-XF™ Differentiation Medium
OsteoMAX-XF™ Differentiation Medium is specially formulated to readily differentiate human MSCs into mature osteocytes as assessed by alizarin red and alkaline phosphatase staining. This proprietary formulation was developed using Plasticell’s CombiCult™, a patented, bead-based, combinatorial technology specially developed for discovery of novel stem cell differentiation protocols.
Learn About Combicult Technology
Application Note: Accelerated Osteocyte Differentiation of Human Mesenchymal Stem Cells in a Novel Xeno-free and Serum-free Osteogenesis Differentiation Media
Features and Benefits:
  • Serum-free, xeno-free, defined medium formulation
  • Rapid, robust, and highly efficient mineralization and bone nodule formation in standard tissue culture system by as early as 7 days of differentiation.
  • Proven to work in 3D BioPrinting applications
“OsteoMAX-XF produces tissue of impressive osteogenic potential, including key bone-specific and mineralization markers. We found we could rapidly fabricate functional, fully human tissue derived from MSCs and the ECM proteins they produce using the NovoGen MMX 3D-Bioprinter”
Dr. Eric Michael David MD JD, Chief Strategy Officer of Organovo
Mineralization kinetics of human bone-marrow derived MSC
Figure 5.Mineralization kinetics of human bone-marrow derived MSC (Product No. SCC034) differentiated in OsteoMAX-XF™ Medium vs. leading competitor osteogenesis media.
Mesenchymal Stem Cell Markers
We offer a variety of positive and negative selection markers, stains, and dyes for MSC imaging and characterization. Browse our Mesenchymal Stem Cell Antibodies here.
Positive Markers: CD105CD73CD90STRO-1
Negative Markers: CD45CD34CD14CD11bCD19HLA-DR
Mesenchymal Stem Cell Related Stains and Dyes:
Alizarin RedOil Red O,  Safranin,  Alcian Blue
Human Mesenchymal Stem Cell Characterization Kit
The Human Mesenchymal Stem Cell Characterization Kit contains a panel of validated positive and negative selection markers for the characterization of the mesenchymal stem cell population in human samples. The kit contains positive antibodies directed against CD44, CD90, STRO-1, and CD146. In addition, two specific hematopoietic antibodies CD14 and CD19.
Immunocytochemical staining of cultured human bone marrow-derived mesenchymal stem cells stained
Figure 6.Immunocytochemical staining of cultured human bone marrow-derived mesenchymal stem cells stained with antibodies provided in the kit. Nuclei of the cells were visualized with DAPI (blue). CD19 and CD14 staining were not present on human mesenchymal stem cells.
Sorry, an unexpected error has occurred
Network error: Failed to fetch