Merck
HomeSmall Molecule HPLCRP Amide C16 HPLC Columns
RP Amide C16 HPLC Columns
Discovery® HPLC Columns for Fast LC-MS Analysis
Discovery RP-Amide C16 HPLC Columns
Polar-embedded phase for unique retention and selectivity
  • Excellent peak shape and efficiency
  • Polar group (amide) gives less retention and different selectivity than a C18
  • Not recommended for LC/MS
Properties
Bonded Phase: Palmitamidopropylsilane, endcappedRP-Amide C16 structure
Silica: Spherical, high purity (Fe <20; NA <7; Ca <7;
Ti <1; AI <1; Mg <1ppm)
Particle Size: 5 µM
Pore Size: 180(Å)
Surface Area: 200m2/g
%C: 11%
Coverage: 2-3µmoles/m2
Basic drugs
Antitussive
Ordering Information
5 µM Discovery RP-Amide C16 HPLC Columns
ID(mm)Length(cm)Cat. No.
2.1550500521
2.110569320-U
2.112.5569329-U
2.11550501321
3.05505005-30
3.010569321-U
3.012.5569330-U
3.015505013-30
3.025505064-30
4.05505005-40
4.010569322-U
4.012.5569331-U
4.015505013-40
4.025505064-40
4.65505005
4.610569323-U
4.612.5569332-U
4.615505013
4.625505064
10.025569324-U
21.225569326-U
5 µM Discovery RP-Amide C16 Validation Packs*
ID(mm)Length(cm)Cat. No.
2.1555705-U21
2.110569802-U
2.11555707-U21
4.6555705-U
4.610569803-U
4.61555707-U
4.62555709-U
* Packs include 3 columns, each from a different lof of bonded phase.
2cm Supelguard Cartridges with Discovery RP-Amide C16 Packings
Description2.1mm ID3.0mm ID4.0mm ID ■
5 µM Particle Size
2 Pack
50511059578-U505099
Kit ▲
50510259577-U505080
■ For 4.0mm ID or 4.6mm ID analytical columns
▲ Kit includes cartridge, stand-alone holder, tubing, 2 nuts and ferrules.
How to Contact Supelco
In USA:
Phone: 800-247-6628 or 814-359-3441
fax: 800-325-5052
email: [email protected]
Outside the USA: click here