Analyze Iron (Total) in Plant Material
The Spectroquant® Iron test allows the accurate quantification of iron ions in various waters. All iron ions are reduced to iron(II) ions by ascorbic acid. In a thioglycolate-buffered medium, these react with a triazine derivative to form a red-violet complex that is measured photometrically.
Materials
- Spectroquant® VIS Spectrophotometer Prove 100 (Product No. 173016), or Spectroquant® UV/VIS Spectrophotometer Prove 300 (Product No. 173017), or Spectroquant® UV/VIS Spectrophotometer Prove 600 (Product No. 173018), or Spectroquant® Photometer NOVA 30 (Product No. 109748), or Spectroquant® Photometer NOVA 60 (Product No. 109751), or Spectroquant® Colorimeter Move 100 (Product No. 173632)
- Rectangular cells 10 mm (Product No. 114946), and/or Rectangular cells 20 mm (Product No. 114947), and/or Rectangular cells 50 mm (Product No. 114944 )
- Spectroquant® Iron Cell Test (Product No. 114549)
- Spectroquant® Iron Cell Test* (Product No. 114896)
- Spectroquant® Iron Test (Product No. 114761)
- Spectroquant® Iron Test (Product No. 100796)
- Nitric acid 65 % for analysis (Product No. 100456)
- Sulfuric acid 95 - 97% for analysis (Product No. 100731)
- Perchloric acid 70% for analysis (Product No. 100519)
- Water for analysis (Product No. 116754)
*not compatible with the Spectroquant® Move 100 hand-held colorimeter
Plant Material Sample Preparation
Ground the plant material finely and pass through a sieve (mesh 0.4 - 0.7 mm) and dry the material at 105 °C for 3 hours.
Add 1 g of the dried material to a mixture of 1.5 mL nitric acid 65 %, 0.5 mL perchloric acid 70 %, 1 mL sulfuric acid 95 - 97 % and heat until white fumes are envolved (fume cupboard). After cooling, dissolve the residue in water for analysis and fill to the mark in a 100-mL standard volumetric flask and mix well.
Analysis
Determine iron content with the above mentioned test kits.
Calculation
Iron (total) content in mg/kg Fe = | Analysis value in mg/L Fe x 100 |
Weight of sample in g |