Merck
All Photos(1)
Key Documents
PM2547050
Millipore
PTFE Membrane Filter, 2.0 μm Pore Size
Fluoropore®, filter diam. 46.2 mm, hydrophobic
Synonym(s):
PTFE for PM2.5 Particle Monitoring, PTFE, hydrophobic, 2.0 µm, 47 mm, white, plain, polypropylene ring, sequential serial number
Sign Into View Organizational & Contract Pricing
Select a Size
50 PKG
$1,050.00
$1,050.00
Estimated to ship on13 April 2025
Select a Size
Change View
50 PKG
$1,050.00
About This Item
UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
$1,050.00
Estimated to ship on13 April 2025
material
PTFE membrane
plain filter
white filter
Quality Level
sterility
non-sterile
feature
hydrophobic
manufacturer/tradename
Fluoropore®
Millipore
filter diam.
46.2 mm
thickness
40 μm
matrix
Fluoropore®
pore size
2.0 μm pore size
shipped in
ambient
General description
Filter Type: Screen filter
Particulates and contaminants in the air affect human capital, the most precious natural resource. Recognizing the need to protect this resource, industries, governments and regulatory agencies around the world monitor ambient air. Millipore′s PTFE Membrane for PM2.5 Particle Monitoring has been developed using US EPA test standards.
Application
PM 2.5 particle monitoring
Legal Information
Fluoropore is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Articles
Considerations in filter selection for particulate matter sampling and measurement for PM2.5 compliance in air quality analysis.
Questions
Reviews
No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service