HomeSolid Phase Microextraction (SPME)GC/MS Analysis of Ethanol in Low Alcohol Beer on SUPELCOWAX® 10 after Headspace SPME using a 100 µm PDMS Fiber
GC/MS Analysis of Ethanol in Low Alcohol Beer on SUPELCOWAX® 10 after Headspace SPME using a 100 µm PDMS Fiber
Conditions | |
---|---|
Sample Preparation | Sample equilibrated prior to extraction for 7 min at 40 °C, with agitation at 250 rpm |
Sample/Matrix | 4 mL of diluted low alcohol beer sample in 10 mL headspace vial |
Desorption Process | 250 °C, 3 min, split 10:1 |
Extraction | Headspace at 40C for 2 min, agitation at 250 rpm |
SPME fiber | Pk/3 SPME Assy, 100 µm, PDMS FS 24 ga Auto (Red) (57301) |
Column | SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 µm (24079) |
Oven | 40 °C (5 min), 8 °C/min to 70 °C, 20 °C/min to 250 °C, held for 5 min |
Carrier Gas | Helium, 1 mL/min constant flow |
Injection | Split, 10:1 |
Liner | 0.75 mm I.D. SPME |
Inj. Temp. | 250 °C |
Detector | MSD |
MSD interface | 250 °C |
Scan Range | m/z 25-300 |
Description | |
---|---|
Analysis Note | Traditional methods for determination of alcohol in beverages include specific gravity and gas chromatography (GC). Specific gravity may not be able to accurately measure levels in beverages with low alcohol content, below 1%. GC is a better approach because it does not have this limitation. However, direct injection of samples containing sugars or other nonvolatile matrix components can foul the GC instrument. This drawback of GC can be overcome by sampling the headspace over the sample using solid phase microextraction (SPME), The method shown here was developed specifically for the determination of ethanol in low alcohol content samples, and was used to analyze a reference material of low alcohol beer. The sample had a certified value of 0.505% alcohol by volume (% ABV). Using headspace SPME, the average value measured was 0.495% ABV, representing 97% accuracy. |
Featured Industry | Food and Beverages |
Legal Information | SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC |
Other Notes | SPME Sample pretreatment: Dilute 400 uL of sample to 4 mL with 0.1M sodium phosphate buffer ( pH 7) containing 25% NaCl |
Suitability | Application for SPME, application for GC/MS |
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch