LC-MS Analysis of Carotene Compounds on Ascentis® RP-Amide
![LC-MS Analysis of Carotene Compounds on Ascentis® RP-Amide application for HPLC](/deepweb/assets/sigmaaldrich/product/images/402/831/ebd4ce3d-0a05-4a3e-8461-6760a345843d/800/ebd4ce3d-0a05-4a3e-8461-6760a345843d.jpg)
Materials
analytical column
Product No.
Description
Pricing
standard
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 2.1 mm I.D., 5 μm particles (565305-U)
mobile phase
acetonitrile
flow rate
0.2 mL/min
column temp.
35 °C
detector
MS, APCI(+), SIM
injection
5 μL
sample
10 mg/L in acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany