Merck
HPLC Analysis of Yohimbe Alkaloids on Ascentis® Express F5, HILIC Mode
HPLC Analysis of Yohimbe Alkaloids on Ascentis® Express F5, HILIC Mode application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express F5, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 3 cm × 3 mm
CONDITIONS
column
Ascentis Express F5, 3 cm x 3.0 mm I.D., 2.7 μm particles (53574-U)
mobile phase
2 mM ammonium acetate in 10:90 water:acetonitrile, pH to 6.0 with acetic acid
flow rate
0.5 mL/min
pressure
1100 psi (76 bar)
column temp.
35 °C
detector
UV, 275 nm
injection
2 μL
sample
100 ug/mL each in methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany