HPLC Analysis of Xylo-oligosaccharide on Proteomix® SAX-NP5
![HPLC Analysis of Xylo-oligosaccharide on Proteomix® SAX-NP5 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/422/735/ffe88aed-0046-466b-94dd-c53cbd7ac583/800/ffe88aed-0046-466b-94dd-c53cbd7ac583.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Proteomix SAX-NP5, 15 cm x 4.6 mm I.D., 5 μm particles (Z777228)
mobile phase
(A) 0.05% ammonium hydroxide in acetonitrile (B) 0.05% ammonium hydroxide in water
gradient
15-60% B (20 min)
flow rate
0.7 mL/min
pressure
1015 psi (70 bar)
temp.
35 °C
detector
ELSD
injection
10 μL
sample
Xylose oligosaccharides
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies