Merck
HPLC Analysis of Triacylglycerols in Soybean Oil on Discovery® HS C18 after Sample Prep using Discovery® DSC-Si (Propionitrile Mobile Phase, ELSD Detection)
HPLC Analysis of Triacylglycerols in Soybean Oil on Discovery® HS C18 after Sample Prep using Discovery® DSC-Si (Propionitrile Mobile Phase, ELSD Detection) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® HS C18 HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
SPE tube or plate
Product No.
Description
Pricing
Discovery® DSC-Si SPE Tube
bed wt. 1 g, volume 6 mL, pk of 30
mobile phase component
Product No.
Description
Pricing
Propionitrile
99%
standard
Product No.
Description
Pricing
Glyceryl trioleate
≥99%
Glyceryl trilinoleate
≥98% (TLC), liquid
Soybean oil
analytical standard
CONDITIONS
condition
6 mL hexane
SPE tube/cartridge
Discovery DSC-Si, 1 gm/6 mL (52656-U)
sample addition
0.12 g oil in 0.5 mL hexane
elution
10 mL hexane-diethyl ether 87-13 (v/v)
eluate post-treatment
vaporation to dryness, reconstituted into 2 mL acetone
column
Discovery HS C18, 25 cm x 4.6 mm I.D., 5 μm particles (568523-U)
mobile phase
propionitrile
flow rate
1.5 mL/min
pressure
1117 psi (77 bar)
column temp.
30 °C
detector
ELSD
injection
5 μL
Description
Analysis Note
Soybean oil was cleaned using Silica SPE and analyzed as 5% sample in acetone (w/v).
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany