HPLC Analysis of Steroids on Ascentis® Express F5, Acetonitrile Mobile Phase
![HPLC Analysis of Steroids on Ascentis® Express F5, Acetonitrile Mobile Phase application for HPLC](/deepweb/assets/sigmaaldrich/product/images/173/115/32ec9086-b6eb-4330-af70-5b889e8767b3/800/32ec9086-b6eb-4330-af70-5b889e8767b3.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53569-U)
mobile phase
[A] water; [B] acetonitrile; (70:30, A:B)
flow rate
0.2 mL/min
column temp.
35 °C
detector
UV, 254 nm
injection
2 μL
sample
50 mg/L in 75:25, water:methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany