Merck
HPLC Analysis of Steroidal Glycosides on Ascentis® Phenyl
HPLC Analysis of Steroidal Glycosides on Ascentis® Phenyl application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Phenyl HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 μm particles (581616-U)
mobile phase
[A] 10 mM ammonium formate (pH 3 with formic acid); [B] methanol
flow rate
1.25 mL/min
column temp.
35 °C
detector
ELSD at 45 °C, gain 6, 3.5 bar N2
injection
20 μL
sample
as indicated in mobile phase A
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany