Merck
HPLC Analysis of Proteins on BIOshell A400 Protein C18
HPLC Analysis of Proteins on BIOshell A400 Protein C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
BIOshell A400 Protein C18 Column
L × I.D. 10 cm × 2.1 mm, 3.4 μm particle size
standard
Product No.
Description
Pricing
β-Galactosidase from Escherichia coli
For use as a marker in SDS-PAGE
Albumin from chicken egg white
For use as a marker in SDS-PAGE
Lysozyme from chicken egg white
powder or granules, ≥39,000 units/mg protein
Conalbumin from chicken egg white
Substantially iron-free
Haptoglobin Human, Phenotype 1-1
98-100%, essentially salt-free, lyophilized powder
Trypsin Inhibitor from Glycine max (soybean)
BioUltra, lyophilized powder, ≥95% (Kunitz inhibitor, SDS-PAGE)
CONDITIONS
column
BIOShell A400 C18, 10 cm x 2.1 mm I.D., 3.4 μm particles (67463-U)
mobile phase
[A] (water, 0.1% TFA); (acetonitrile, 0.08% TFA), 75:25; [B] (water, 0.1% TFA); (acetonitrile, 0.08% TFA), 25:75
gradient
20 to 100% B in 8 min
flow rate
0.3 mL/min
pressure
2480 psi total pressure (193 bar)
column temp.
30 °C
detector
UV, 215 nm
injection
2 μL
sample
lysozyme, 0.06; haptoglobulin, 0.06; conalbumin, 0.12; tryspin inhibitor, 0.18; glutamate dehydrogenase, (0.18; ß-galactosidase, 0.12; serum albumin chicken, 0.12 g/L in mobile phase A)
Description
Analysis Note
Analysis of proteins of various hydrophobicity on BIOshell A400 Protein C18 to compare the selectivity and retention times.
Legal Information
null