Merck
HPLC Analysis of Luciferin Enantiomers on CHIRALPAK® AGP
HPLC Analysis of Luciferin Enantiomers on CHIRALPAK® AGP application for HPLC
Materials
analytical column
Product No.
Description
Pricing
CHIRALPAK® AGP HPLC Column
5 μm particle size, L × I.D. 10 cm × 4 mm
standard
Product No.
Description
Pricing
D-Luciferin sodium salt
lyophilized powder, synthetic
D-Luciferin
synthetic, BioXtra, ≥99% (HPLC)
D-Luciferin
synthetic
CONDITIONS
column
CHIRALPAK AGP, 10 cm x 4 mm I.D., 5 μm particles (58150AST)
mobile phase
10 mM sodium phosphate, pH 6.0
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 225 nm
sample
luciferin
Description
Legal Information
CHIRALPAK is a registered trademark of Daicel Corp.