HPLC Analysis of Isoleucine Enantiomers on Astec® CHIROBIOTIC® T (23 °C )
![HPLC Analysis of Isoleucine Enantiomers on Astec® CHIROBIOTIC® T (23 °C ) application for HPLC](/deepweb/assets/sigmaaldrich/product/images/344/332/de39cf41-87a9-44e7-9724-5fb3a270c242/800/de39cf41-87a9-44e7-9724-5fb3a270c242.jpg)
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] ethanol; (70:30, A:B)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 205 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC